06999
Conductivity Standard
conductivity 500 μS/cm at 25 °C, ≤2.5% KCl (T)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
≤2.5% KCl (T)
conductivity
495.0-505.0 μS/cm at 25 °C
density
0.995-1.014 g/cm3 at 20 °C
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
single component solution
Looking for similar products? Visit Product Comparison Guide
General description
Conductivity standard can generally be used for the calibration of conductivity measurements. The conductivity calibration standard solution used for conductivity measurements can be sodium chloride or potassium chloride solution of a definite concentration.
Application
- UPLC-TQD-MS/MS Method Validation: A study developed an ultra-performance liquid chromatography-tandem quadrupole mass spectrometry (UPLC-TQD-MS/MS) method, utilizing conductivity standards for precise quality control of alkaloid content in food and dietary supplements containing Lepidium meyenii (Maca). This application emphasizes the role of conductivity standards in ensuring pharmaceutical grade accuracy and reliability in biotech research (Le NT et al., 2024).
- Chemical Constituents Analysis: Research on the chemical constituents of Gastrodia tuber hallimasch powder was facilitated by the use of high-precision conductivity standards in UHPLC-Q-Orbitrap HRMS and HPLC analysis, underscoring their critical role in maintaining traceability and accuracy in biotechnological applications (Wu Y et al., 2024).
- Pharmaceutical Applications of Castanea sativa: An investigation into the potential food and pharmaceutical applications of Castanea sativa wood extract highlighted the use of certified conductivity standards to assess the extract′s quality. This research demonstrates the utility of conductivity standards in developing biochemically traceable solutions for scientific research (Žitek Makoter T et al., 2024).
- Biopharmaceutics Modeling: The summary report from a 2023 workshop on Physiologically Based Biopharmaceutics Modeling (PBBM) discussed the integration of conductivity standards in biopharmaceutics, indicating their essential role in enhancing drug product quality and regulatory compliance in the pharma and biotech sectors (Mackie C et al., 2024).
standard
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - STOT RE 2 Oral
Target Organs
Liver,Kidney
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Analysis and Analyzers, Volume 2 (2016)
Volunteer stream monitoring a methods manual null
Analysis and Analyzers, Volume 2 (2016)
Volunteer stream monitoring a methods manual null
Volunteer stream monitoring a methods manual null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service