Skip to Content
Merck
All Photos(1)

Key Documents

06444

Supelco

Amberlite XAD4 polymeric adsorbent

20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

product name

Amberlite XAD4,

form

beads

autoignition temp.

800 °F

technique(s)

LPLC: suitable

surface area

750 m2/g

loss

~55% loss on drying, 110 °C

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.98 mL/g pore volume
100 Å mean pore size

density

1.02 g/mL (true wet)(lit.)
1.08 g/mL (skeletal)(lit.)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Amberlite XAD-4 has been used in the clean-up procedure prior to high performance liquid chromatography-evaporative laser scattering detector (HPLC-ELSD) analysis of mycotoxin fumonisins in corn products. It has also been used as a solid sorbent to trap imidacloprid and its metabolite 6-chloronicotinic acid from greenhouse air prior to their determination by HPLC coupled to diode array detector (DAD).
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.

Other Notes

Two particle sizes are available:
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of mycotoxin fumonisins in corn products by high-performance liquid chromatography coupled with evaporative light scattering detection.
Wang J, et al.
Food Chemistry, 107(2), 970-976 (2008)
Antoine P Trzcinski et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 46(13), 1539-1548 (2011-10-14)
In this study, various methods were compared to reduce the Chemical Oxygen Demand (COD) content of stabilised leachate from a Submerged Anaerobic Membrane Bioreactor (SAMBR). It was found that Powdered Activated Carbon (PAC) resulted in greater COD removals (84 %)
A S Ghatbandhe et al.
Journal of environmental science & engineering, 50(2), 163-168 (2009-03-20)
Adsorption equilibrium, kinetics and thermodynamics of 2,4-dichlorophenol (2,4-DCP), one of the most commonly used chlorophenol, onto bituminous coal based Filtrasorb-400 grade granular activated carbon, were studied in aqueous solution in a batch system with respect to temperature. Uptake capacity of
Muhammad Afzal Kamboh et al.
Journal of hazardous materials, 172(1), 234-239 (2009-07-31)
The present study describes a novel synthetic method for the immobilization of calix[4]arene (II) onto the surface of modified Amberlite XAD-4 resin (4), which does not require the derivatization of calixarene moiety. The novel calix[4]arene based resin (C4 resin) 5
Valfredo Azevedo Lemos et al.
Environmental monitoring and assessment, 171(1-4), 163-169 (2009-12-17)
Amberlite XAD-4 resin functionalized with β-nitroso-α-naphthol was applied to an online system for the preconcentration and determination of uranium. U (VI) ions were retained on the minicolumn at an appropriate pH and then desorbed with acid solution. The amount of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service