Skip to Content

05040

Agar

powder, suitable for microbiology

Synonym(s):

Agar-agar, Gum agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
eCl@ss:
42040102
UNSPSC Code:
41106212
NACRES:
NA.74
EC Number:
232-658-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg, pkg of 100 g, pkg of 250 g

storage condition

(Tightly closed)

ign. residue

≤6%

loss

≤20% loss on drying

pH

5-8 (50 °C, 1.5% in H2O, 100 °C/15 min.)

transition temp

gel point ~35 °C (1.5% solution)

gel strength

>300 g/cm2 (1.5% gel)

solubility

H2O: 1.5% at 70 °C, clear to slightly hazy (100 °C/15 min.)

application(s)

agriculture, microbiology

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

General description

Agar is widely used in microbiology as a solidifying agent in microbial culture media. It is a polysaccharide derived from seaweed and is primarily composed of agarose. Agar is a mostly inert substance, which makes it an ideal material for microbial culture media because it doesn’t interfere with the growth of microorganisms. Agar can solidify media, allowing microorganisms to grow in a solid environment to promote the formation of distinct colonies and enable selective isolation of specific microorganism. It is also used to prepare selective and differential media, which can provide additional nutrients or other chemical substances to promote or inhibit the growth of specific microorganisms selectively. Agar is sterilized by autoclaving and then poured into Petri dishes to cool and solidify, forming the surface for microbial growth.

Application

Agar is commonly used as a solidifying agent in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.


Still not finding the right product?

Explore all of our products under Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library