Skip to Content
Merck
All Photos(1)

Key Documents

04991

Supelco

2,6-Lutidine

analytical standard

Synonym(s):

2,6-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
105690
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.497 (lit.)
n20/D 1.499

bp

143-145 °C (lit.)

mp

−6 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1cccc(C)n1

InChI

1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3

InChI key

OISVCGZHLKNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F

Flash Point(C)

32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Arceo et al.
Nature chemistry, 5(9), 750-756 (2013-08-24)
Asymmetric catalytic variants of sunlight-driven photochemical processes hold extraordinary potential for the sustainable preparation of chiral molecules. However, the involvement of short-lived electronically excited states inherent to any photochemical reaction makes it challenging for a chiral catalyst to dictate the
Jun Gao et al.
Journal of hazardous materials, 130(1-2), 141-147 (2005-10-26)
The homogeneous catalytic N-oxidation of two picolines and two lutidines by hydrogen peroxide has been studied calorimetrically using a heat flow and power compensation SIMULAR reaction calorimeter. The objective of this work was to extend a previously developed kinetic model
Kathryn A White et al.
Journal of colloid and interface science, 359(1), 126-135 (2011-04-22)
Using a system of modified silica particles and mixtures of water and 2,6-lutidine to form particle-stabilized emulsions, we show that subtle alterations to the hydration of the particle surface can cause major shifts in emulsion structure. We use fluorescence confocal
Cyrille Costentin et al.
Journal of the American Chemical Society, 132(29), 10142-10147 (2010-07-06)
The effect of base pairing by cytosine on the electrochemical oxidation of guanine is examined by means of cyclic voltammetry on carefully purified reactants in a solvent, CHCl(3), which strongly favors the formation of an H-bonded pair. The thermodynamics and
Wensheng Yu et al.
Organic letters, 6(19), 3217-3219 (2004-09-10)
[reaction: see text] Oxidative cleavage of olefins by OsO(4)-NaIO(4) sometimes suffers from low yields due to the formation of side products. It is found that the addition of 2,6-lutidine can suppress the side reactions and dramatically improve the yield of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service