Skip to Content
Merck

04739

Indoxyl Strips

suitable for microbiology

Synonym(s):

Acetoxyindol Strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171621
Agency:
according to ISO 10272-1:2017, according to ISO 10272-2:2017
Suitability:
Moraxella spp., Campylobacter spp.
Application(s):
clinical testing
food and beverages, microbiology
Technique(s):
microbe id | specific enzyme detection: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

according to ISO 10272-1:2017, according to ISO 10272-2:2017

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages, microbiology

storage temp.

2-8°C

suitability

Moraxella spp., Campylobacter spp.

Application

Diagnostic test for the rapid detection of the acetate esterase activity of microorganisms. The test is based on the reaction of free indoxyl groups with oxygen, which results in a colour change. Acetate esterase activity is present e.g. in Campylobacter spp., and in Moraxella (Branhamella) catarrhalis. The test is suitable for screening examinations and for the suspect microbial colonies identification. The result of the reaction is read after 3-5 min.

Packaging

100 test strips

Other Notes

The kit contains plastic strips with an active zone saturated with 3-acetoxyindol, which is a substrate for acetate esterase.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library