Skip to Content
Merck

04397

Conjugated (10E,12Z)-Linoleic acid solution

~100 mg/mL in ethanol, analytical standard

Synonym(s):

(10E,12Z)-10,12-Octadecadienoic acid, 10E,Z12-CLA, Linoleic acid (10-trans, 12-cis)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

grade

analytical standard

assay

≥96.0% (HPLC)

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

~100 mg/mL in ethanol

format

single component solution

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

CCCCC/C=C\C=C/CCCCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-9H,2-5,10-17H2,1H3,(H,19,20)/b7-6-,9-8+

InChI key

GKJZMAHZJGSBKD-NMMTYZSQSA-N



pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

57.2 °F

flash_point_c

14 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library