Skip to Content
Merck

04229

Mettler-Toledo Calibration substance ME 30130610, Sodium methanesulfonate

traceable to primary standards (LGC)

Synonym(s):

Sodium methanesulfonate, Methanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3SO3Na
CAS Number:
Molecular Weight:
118.09
UNSPSC Code:
12352302
NACRES:
NA.24
PubChem Substance ID:
EC Number:
219-203-2
Beilstein/REAXYS Number:
3915209
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages
pharmaceutical

SMILES string

[Na+].CS([O-])(=O)=O

InChI

1S/CH4O3S.Na/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

KKVTYAVXTDIPAP-UHFFFAOYSA-M

General description

Mettler-Toledo calibration substance ME 30130610, sodium methanesulfonate is an analytical standard, suitable for the regular calibration of Mettler-Toledo melting point instrument. The melting point valueequals a mean of 6 to 12 measurements with aMettler-Toledo MP90 Excellence instrument that is calibrated withprimary standards. The melting point is validated by Capillary method according to European Pharmacopeia (2.2.14.)

Application

Sodium methanesulfonate Mettler-Toledo calibration substance ME 30130610 is used to calibrate Mettler-Toledo melting point instruments.

Features and Benefits

  • Melting point calibration standard traceable to a primary standard (LGC)
  • Melting point measured in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point analyses, respectively
  • Provided with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library