Skip to Content
Merck

Skip To

04070

Alkane standard solution C8-C20

analytical standard, contains C8-C20, ~40 mg/L each, in hexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
EC Number:
203-777-6
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

refractive index

n20/D 1.375

application(s)

petroleum

format

multi-component solution

Application

Alkane standard solution C8-C20 has been used as standard in the measurement of retention indices for the identification of alkanes co-existing in essential oils using gas chromatography coupled with mass spectrometry.
Alkane standard solution C8-C20 may also be used as an analytical standard for measurement of linear retention indices (LRI) in the identification of volatile components in honey samples using gas chromatography-mass spectrometry (GC/MS).

Other Notes

contains C8, C9, C10, C11, C12, C13, C14, C15, C16, C17, C18, C19, C20; ~40 mg/l each, in hexane

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4710804071909505
description

analytical standard, contains C8-C20, ~40 mg/L each, in hexane

description

certified reference material, 2 % (w/w) each component in octane

description

analytical standard

description

Mixture of Diastereomers

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

application(s)

-

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

-

grade

analytical standard

grade

TraceCERT®, certified reference material

grade

analytical standard

grade

-

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

-

refractive index

n20/D 1.375

refractive index

-

refractive index

n20/D 1.497

refractive index

-


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. I cant see a list of the CAS numbers for the added alkanes. Do you have it?

    1 answer
    1. This standard mixture contains octane (CAS No. 111-65-9), nonane (CAS No. 111-84-2), decane (CAS No. 124-18-5), undecane (CAS No. 1120-21-4), dodecane (CAS No. 112-40-3), tridecane (CAS No. 629-50-5), tetradecane (CAS No. 629-59-4), pentadecane (CAS No. 629-62-9), hexadecane (CAS No. 544-76-3), heptadecane (CAS No. 629-78-7), octadecane (CAS No. 593-45-3), nonadecane (CAS No. 629-92-5), and icosane (CAS No. 112-95-8).

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Are these all n-alkanes? So for example, is the C8 only octane or is it a mixture of isomers? 2-methylheptane, 3-methylheptane, etc. Thank you!

    1 answer
    1. This product contains ~40 mg/l of each C8, C9, C10, C11, C12, C13, C14, C15, C16, C17, C18, C19 and C20 in hexane.

      Helpful?

Reviews

No rating value

Active Filters