Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3(CH2)5COCH3
CAS Number:
Molecular Weight:
128.21
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-837-1
Beilstein/REAXYS Number:
635843
MDL number:
grade
analytical standard
Quality Level
assay
≥99.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
refractive index
n20/D 1.416
bp
173 °C (lit.)
mp
−16 °C (lit.)
density
0.819 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCCCCCC(C)=O
InChI
1S/C8H16O/c1-3-4-5-6-7-8(2)9/h3-7H2,1-2H3
InChI key
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
General description
2-Octanone is a volatile compound reported to be found in blue cheese, acerola fruit and blue crab.
Application
2-Octanone may be used as an analytical reference standard for the quantification of the analyte in sugar-cane spirits and rum samples and whey protein concentrate using chromatography based techniques.
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Still not finding the right product?
Explore all of our products under 2-Octanone
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
132.8 °F - closed cup
flash_point_c
56 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Volatile component change in whey protein concentrate during storage investigated by headspace solid-phase microextraction gas chromatography
Javidipour I and Qian CM
Dairy science & technology, 88(1), 95-104 (2008)
Jian Hu et al.
Biotechnology letters, 28(14), 1115-1119 (2006-06-24)
An aqueous-organic biphasic system was established and used with whole cells of Oenococcus oeni to reduce 2-octanone to (R)-2-octanol. The conversion reached 99% when the Tris/borate buffer was increased from 50 mM to 300 mM in the aqueous phase. In
J S Hosler et al.
The Journal of experimental biology, 203(Pt 18), 2797-2806 (2000-08-23)
Recent studies of olfactory blocking have revealed that binary odorant mixtures are not always processed as though they give rise to mixture-unique configural properties. When animals are conditioned to one odorant (A) and then conditioned to a mixture of that
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 138150-25G | 04061831828320 |
| 138150-1G | 04061825582962 |
| 138150-5G | 04061831809794 |
| 02479-5ML | 04061837239755 |
| 02479-1ML | 04061838617354 |

