Skip to Content
Merck

02193

Monononadecanoin

analytical standard

Synonym(s):

Glyceryl monononadecanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H44O4
CAS Number:
Molecular Weight:
372.58
UNSPSC Code:
85151701
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCC(=O)OCC(O)CO.CCCCCCCCCCCCCCCCCCC(=O)OC(CO)CO

InChI

1S/2C22H44O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(25)26-20-21(24)19-23;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(25)26-21(19-23)20-24/h2*21,23-24H,2-20H2,1H3

InChI key

SNBRFGSCPLJOMA-UHFFFAOYSA-N

Application

Monononadecanoin may be used as a reference standard for the separation and isolation of neutral lipid from a lipid mixture into individual class, in high yield and purity, by using bonded phase columns.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library