Skip to Content
Merck
All Photos(1)

Key Documents

02010595

Valerenic acid

primary reference standard

Synonym(s):

(2E)-3-[(4S,7R,7aR)-2,4,5,6,7,7a-Hexadydro-3,7-dimethyl- 1H-inden-4-yl]-2-methyl-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
Beilstein:
3138020
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

SMILES string

[H][C@]12CCC(C)=C1[C@@H](CC[C@H]2C)\C=C(/C)C(O)=O

InChI

1S/C15H22O2/c1-9-4-6-12(8-11(3)15(16)17)14-10(2)5-7-13(9)14/h8-9,12-13H,4-7H2,1-3H3,(H,16,17)/b11-8+/t9-,12+,13-/m1/s1

InChI key

FEBNTWHYQKGEIQ-SUKRRCERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: valeriana

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Khom et al.
British journal of pharmacology, 161(1), 65-78 (2010-08-20)
Subunit-specific modulators of gamma-aminobutyric acid (GABA) type A (GABA(A)) receptors can help to assess the physiological function of receptors with different subunit composition and also provide the basis for the development of new drugs. Valerenic acid (VA) was recently identified
Andres Navarrete et al.
Journal of AOAC International, 89(1), 8-15 (2006-03-04)
The roots and rhizomes of various valeriana species are currently used as a sleeping aid or mild sedative. A liquid chromatography method has been developed that permits the analysis of chlorogenic acid, lignans, flavonoids, valerenic acids, and valpotrates in various
Jürgen Ramharter et al.
Organic letters, 11(5), 1151-1153 (2009-01-31)
The first total synthesis of the sesquiterpenoid valerenic acid, a constituent of Valeriana officinalis, is described. The compound is a potent modulator of the GABA(A) receptor and may thus be useful in the treatment of various dysfunctions of the central
Chethan Sampath et al.
Planta medica, 78(6), 575-581 (2012-03-14)
Valerenic acid (VA), a sesquiterpenoid, is one of the major secondary bioactive metabolites of VALERIANA OFFICINALIS L. Until now IN VIVO studies on the absorption, bioavailability, disposition, and metabolism of VA are limited. We established and validated an LC-MS/MS assay
Winfried Neuhaus et al.
Planta medica, 74(11), 1338-1344 (2008-08-16)
The roots and rhizome of Valeriana officinalis L . s. l. are therapeutically used for their sedative and sleep-enhancing effects. Some of the active compounds found in commonly used extracts are the sesquiterpenic acids, especially valerenic acid, which was recently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service