Skip to Content
Merck
All Photos(1)

Documents

01840

Sigma-Aldrich

Adenine monohydrochloride

≥99.0% (HPLC)

Synonym(s):

Adenine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5 · HCl
CAS Number:
Molecular Weight:
171.59
Beilstein:
4009585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (HPLC)

loss

≤5% loss on drying

mp

~285 °C (dec.)

SMILES string

Cl[H].Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5.ClH/c6-4-3-5(9-1-7-3)10-2-8-4;/h1-2H,(H3,6,7,8,9,10);1H

InChI key

UQVDQSWZQXDUJB-UHFFFAOYSA-N

Application

Adenine monohydrochloride can be used in designing novel bioactive adenine derivatives and adenine-based polyimide/nano-silica hybrid films.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and DNA binding of new adenine derivatives incorporating 5-((Alkylthio)-1,3,4-oxadiazol-2-yl) methyl moiety
Y?lmaz Y, et al.
Medicinal Chemistry Research, 27(8), 2027-2033 (2018)
Preparation and characterization of adenine-based polyimide/nano-silica hybrid films
Lu Z, et al.
European Polymer Journal, 102, 209-218 (2018)
Synthesis of triazolylmethyl-linked nucleoside analogs via combination of azidofuranoses with propargylated nucleobases and study on their cytotoxicity
Halay E, et al.
Chemistry of Heterocyclic Compounds, 1-9 (2018)
Shi Chen et al.
Nanoscale, 12(5), 3302-3307 (2020-01-24)
Self-assembly of nucleic acid nanostructures is driven by selective association of oligonucleotide modules through base pairing between complementary sequences. Herein, we report the development of RNA-DNA hybrid nanoshapes that conditionally assemble under the control of an adenosine ligand. The design

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service