Skip to Content
Merck

01422

Melting point standard 79-81°C

analytical standard

Synonym(s):

Naphthalene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
UNSPSC Code:
20121904
NACRES:
NA.24
PubChem Substance ID:
EC Number:
202-049-5
Beilstein/REAXYS Number:
1421310
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

4.4 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

autoignition temp.

978 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.9 %

bp

218 °C (lit.)

mp

79-81 °C (±0.3°C), 80-82 °C (lit.)

application(s)

food and beverages
pharmaceutical

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

General description

Melting point standard 79-81 °C, naphthalene is a non-polar hydrocarbon for use in the characterization of pure chemicals and pharmaceutical compounds. The melting point value is determined with an average of 6 to 12 measurements using a Büchi B-545 instrument that is calibrated against primary standards.

Application

Naphthalene melting point standard 79-81 °C is an analytical standard used to calibrate melting point instruments, operating in the thermodynamic mode of analysis. It′s also applicable in R&D labs for chemical quality control (QC) and quality assurance (QA) to identify solid crystalline substances and to check their purity.
Additionally, this mp standard is also used as a temperature calibration standard in thermal studies.

Features and Benefits

  • Melting point calibration standard traceable to primary standards (LGC, London)
  • Grade: Analytical Standard
  • Standard deviation up to ± 0.3 °C
  • Provided with certificates of analysis and safety data sheet



signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

173.3 °F - closed cup

flash_point_c

78.5 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library