Skip to Content

01419

1,2-Propanediamine-1,2,3-13C3,1-15N hydrochloride

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C3H1015NN · 2HCl
Molecular Weight:
151.02
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

isotopic purity

≥99.0%

product line

PESTANAL®

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

−20°C

SMILES string

[13CH3][13CH](N)[13CH2][15NH2]

InChI

1S/C3H10N2/c1-3(5)2-4/h3H,2,4-5H2,1H3/i1+1,2+1,3+1,4+1

InChI key

AOHJOMMDDJHIJH-JCDJMFQYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library