Skip to Content
Merck
All Photos(1)

Key Documents

0120569

Millipore

mEHEC Broth in bags

For selective enrichment of Enterohemorrhagic E. coli O157:H7 and Salmonella in selected foods, suitable for microbiology

Synonym(s):

Modified Enterohemorrhagic Escherichia coli Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

form

liquid

shelf life

limited shelf life, expiry date on the label

feature

ready-to-use

application(s)

food and beverages
industrial qc
pathogen testing

microbiology

suitability

enterovirulent E. coli (EEC)

General description

mEHEC Broth in bags is used for selective enrichment of Salmonella, Enterohemorrhagic E. coli during bacteriological examination of water, milk and foods. The media contains tryptone that provides nitrogenous and carbonaceous compounds, amino acids and other essential growth nutrients. It contains buffereing agents and selective substances, but the exact composition is proprietary and cannot not be disclosed.

Application

mEHEC Broth in bags is used for selective enrichment of Salmonella, Enterohemorrhagic E.coli O157:H7, and non-O157 Shiga toxin-producing E. coli in selected foods. Specially validated for use with Assurance GDS®, TRANSIA® Plate, TRANSIA® AG, and VIP® Gold.

Components

mEHEC Media contains following nutrients sources: casein digest, yeast digest, carbohydrates, and salts. The exact composition of this proprietary media cannot be disclosed.

Legal Information

Assurance GDS is a registered trademark of BioControl Systems, Inc.
TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany
VIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service