Skip to Content
Merck
All Photos(1)

Key Documents

00320188

(−)-α-Bisabolol

primary reference standard

Synonym(s):

(−)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

storage temp.

−20°C

SMILES string

C\C(C)=C\CC[C@](C)(O)[C@H]1CCC(C)=CC1

InChI

1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1

InChI key

RGZSQWQPBWRIAQ-CABCVRRESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: angelica

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Tschiggerl et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 67(2), 129-135 (2012-03-14)
Chamomile (German Chamomile, Matricaria recutita L., Asteraceae) is one of the most popular medicinal plants in use as an herbal tea for food purposes and in folk medicine. Qualitative and semi-quantitative analyses of the volatile fraction of chamomile herbal tea
Seyed Mehdi Razavi
Natural product research, 26(22), 2148-2151 (2011-11-26)
Prangos pabularia Lindl. (Apiaceae) is a widespread perennial herb distributed from Russia to Iran, Turkey, India, Central Asia and Caucasian. It has been used in folk medicine from ancient times as a diuretic agent and a treatment for leukoplakia, digestive
Seungbeom Kim et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(10), 2580-2585 (2011-07-21)
Although (-)-α-bisabolol, a natural monocyclic sesquiterpene alcohol, is often used as a cosmetic soothing supplement, little is known about its mechanisms of anti-inflammatory effects. Therefore, this study was designed to investigate anti-inflammatory effects of (-)-α-bisabolol and its mechanisms of action.
Orazio Taglialatela-Scafati et al.
Journal of natural products, 75(3), 453-458 (2012-02-09)
Apart from a large amount (ca. 2.0%) of α-bisabolol β-D-fucopyranoside (2a), the aerial parts of the Mediterranean weed Carthamus glaucus afforded an unusual triglyceride (E-2-crotonyl-1,3-distearolylglycerol, 7), two lipophilic flavonoids (6a,b), and a series of bisabolane fucopyranosides variously acylated on the
Maryam Yousefi et al.
Natural product research, 26(2), 146-151 (2011-08-04)
The chemical composition of the essential oil obtained from the aerial parts of Betonica grandiflora Willd., growing wild in Iran, was analysed by gas chromatography-mass spectrometry for the first time. Overall, 40 volatile components were identified on the basis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service