00090
Acetaldehyde ammonia trimer
≥96.0% (NT)
Synonym(s):
Hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H15N3 · 3H2O
CAS Number:
Molecular Weight:
183.25
Beilstein:
3910586
EC Number:
MDL number:
UNSPSC Code:
12352301
eCl@ss:
39021102
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥96.0% (NT)
form
powder
mp
95-97 °C
storage temp.
2-8°C
SMILES string
O.O.O.CC1NC(C)NC(C)N1
InChI
1S/C6H15N3.3H2O/c1-4-7-5(2)9-6(3)8-4;;;/h4-9H,1-3H3;3*1H2
InChI key
VNJBGAOFYNEMRG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Acetaldehyde ammonia trimer is a reagent used in the synthesis of amines by hydrogenation and as a precursor to the production of ammonia. It can also be used as an organic building block in the synthesis of 1,2-dihydro-1,3-dimethyl-3H-naphth[1.2-e]-m-oxazine by condensing with 2-naphthol.
Other Notes
Structure
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A.T. Nielson et al.
The Journal of Organic Chemistry, 38, 3288-3288 (1973)
Condensation of 2-naphthol with acetaldehyde ammonia.
Burke, William J and Reynolds, Richard J
Journal of the American Chemical Society, 76(5), 1291-1293 (1954)
pH effects in the acetaldehyde-ammonia reaction.
Moioli, Emanuele et al.
Reaction Chemistry & Engineering, 2(3), 382-389 (2017)
Einar Wang Lund
Acta Chemica Scandinavica, 12, 1768-1776 (1958)
Catarina S Silva et al.
Biomacromolecules, 21(12), 4771-4780 (2020-11-26)
Thymic epithelial cells (TECs) are the main regulators of T lymphocyte development and selection, requiring a three-dimensional (3D) environment to properly perform these biological functions. The aim of this work was to develop a 3D culture substrate that allows the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service