Skip to Content
Merck
All Photos(1)

Key Documents

00010

Sigma-Aldrich

Abietic acid

technical, ~75% (GC)

Synonym(s):

7,13-abietadien-18-oic acid, Abietate, L-Abietic acid, Rosin acid, Sylvic acid, sylvic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Beilstein:
2221451
EC Number:
MDL number:
UNSPSC Code:
12352002
eCl@ss:
32150233
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

~75% (GC)

form

solid

optical activity

[α]20/D −85±10°, c = 1% in ethanol

mp

139-142 °C (lit.)
150-165 °C

storage temp.

2-8°C

SMILES string

CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

InChI

1S/C20H30O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h7,12-13,16-17H,5-6,8-11H2,1-4H3,(H,21,22)/t16-,17+,19+,20+/m0/s1

InChI key

RSWGJHLUYNHPMX-ONCXSQPRSA-N

Gene Information

human ... TNF(7124)

Looking for similar products? Visit Product Comparison Guide

Application

Abietic acid can be used as a precursor in the preparation of:       
  • Epoxy and petrochemical curing agents.
  •  Dehydroabietic acid derivatives as potential antitumor, antimycotic, and antiviral agents.

It can also be used as a starting material in the total synthesis of:
  • (−)-triptonide and (−)-triptolide as potent antitumor and immunosuppressant agents       
  • quinone (−)-12-deoxyroyleanone as antileishmanial agent       
  • abietic acid-derived catechol (methyl 11,12-dihydroxyabietate-8,11,13-trien-18-oate, MDTO) as a potential antioxidant agent

Other Notes

Starting material for natural product synthesis; Epoxidation with MCPBA

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Regioselective routes towards 14-hydroxyabietane diterpenes. A formal synthesis of immunosuppressant (−)-triptolide from (+)-abietic acid.
Alvarez-Manzaneda E, et al.
Tetrahedron, 63(45), 11204-11212 (2007)
Rosin-based acid anhydrides as alternatives to petrochemical curing agents.
Liu X, et al.
Green Chemistry, 11(7), 1018-1025 (2009)
G. Buchbauer
Monatshefte fur Chemie / Chemical Monthly, 116, 1345-1345 (1985)
Cytotoxic, immunomodulatory, antimycotic, and antiviral activities of semisynthetic 14-hydroxyabietane derivatives and triptoquinone C-4 epimers
Zapata B, et al.
MedChemComm, 4(9), 1239-1246 (2013)
H. Okawara et al.
Tetrahedron Letters, 23, 1087-1087 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service