Skip to Content
Merck

RAP-RO

Roche

rAPid Alkaline Phosphatase

1 units/μg protein, optimum reaction temp. 37 °C

Synonym(s):

alkaline phosphatase, cloning of dna fragments

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

EC Number:
NACRES:
NA.21
UNSPSC Code:
12352204
Specific activity:
1 units/μg protein
Biological source:
bovine intestine
Recombinant:
expressed in Pichia pastoris
Concentration:
<0.1 % (w/w)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

rAPid Alkaline Phosphatase, 1 units/μg protein, optimum reaction temp. 37 °C

biological source

bovine intestine

recombinant

expressed in Pichia pastoris

form

liquid

specific activity

1 units/μg protein

mol wt

monomer 56  kDa by SDS-PAGE

packaging

pkg of 1,000 U (04898133001 [1 U/μl])
pkg of 5,000 U (04898141001 [1 U/μl])

manufacturer/tradename

Roche

concentration

<0.1 % (w/w)

parameter

37 °C optimum reaction temp.

technique(s)

molecular cloning: suitable

color

colorless

pH

7.6-8.5 (68 °F)

solubility

water: miscible

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Nicking activity, none detected (up to 20 µl enzyme using pBR322-DNA/ 16h / 37°C)
unspecific endonuclease, none detected (up to 20 µl enzyme using Lambda-DNA 16 h/37 °C)

storage temp.

−20°C

Quality Level

Gene Information

bovine ... ALPI(280993)

Related Categories

Analysis Note

All lots of rAPid Alkaline Phosphatase are tested for the absence of deoxyribonucleases and nicking activity according to the current Quality Control procedures.

Application

  • Use rAPid Alkaline Phosphatase in numerous applications, including: Removal of 5′-phosphoryl groups from nucleic acids
  • Preparation of templates for 5′-end labeling
  • Clean-up of PCR products by removal of dNTPs
  • Dephosphorylation of proteins
Note: rAPid Alkaline Phosphatase is very rapidly inactivated by heating at +75 °C for two minutes. No additional purification steps are required after restriction and dephosphorylation. The dephosphorylated DNA can directly be used in a ligation reaction.
Note: rAPid Alkaline Phosphatase is also included in the Rapid DNA Dephos & Ligation Kit, which enables fast and efficient dephosphorylation and ligation of sticky- or blunt-end DNA fragments. With this convenient kit, dephosphorylation of blunt and sticky 5′-overhanging ends is performed in ten minutes at +37 °C and ligation is completed in just five minutes at +15–25 °C.

Biochem/physiol Actions

Alkaline Phosphatase catalyzes the hydrolysis of numerous phosphate esters, such as esters of primary and secondary alcohols, saccharides, cyclic alcohols, phenols and amines. Phosphodiesters do not react. The enzyme hydrolyzes inorganic pyrophosphate. The kinetic properties of the enzyme depend on many factors, such as purity of enzyme, concentration of enzyme in the assay, buffer, pH etc.
Heat inactivation: rAPid Alkaline Phosphatase is inactivated by incubation at 75 °C for two minutes.

Features and Benefits

  • Eliminate contamination risk: Recombinant expression, and reliable and sensitive quality control tests ensure high purity.
  • Replace your current phosphatases with an economical solution: rAPid Alkaline Phosphatase performs equally as well as Shrimp Alkaline Phosphatase or Bovine Alkaline Phosphatase.
  • Eliminate the risk of animal-derived diseases: Expression in Pichia pastoris ensures consistency and safety.
  • Benefit from rapid heat inactivation: rAPid Alkaline Phosphatase is inactivated after only 2 minutes at +75°C.
  • Obtain consistent performance: rAPid Alkaline Phosphatase has excellent stability during storage compared to other alkaline phosphatases.

General description

rAPid Alkaline Phosphatase is an alkaline phosphatase isolated from bovine intestine and supplied as a recombinant enzyme of 56 kD, expressed in the yeast, Pichia Pastoris. The recombinant form ensures consistency and safety. rAPid Alkaline Phosphatase catalyzes the dephosphorylation of 5′ phosphates from DNA and RNA, nucleotides, and proteins. Alkaline phosphatase treatment prevents self-ligation of DNA fragments by removing the 5′-phosphoryl termini required by ligases; this feature is of major importance in cloning strategies to decrease vector background. Unlike calf intestinal phosphatase, rAPid Alkaline Phosphatase is rapidly, completely, and irreversibly inactivated by heat treatment for two minutes at +75 °C. It is therefore, an excellent alternative to Shrimp Alkaline Phosphatase. In addition, the enzyme is active in restriction enzyme buffers. Therefore, restriction enzyme digestion, dephosphorylation, enzyme inactivation, ligation, or 5′-end labeling can be performed without purification steps.

Other Notes

For life science research only. Not for use in diagnostic procedures.
One unit of rAPid Alkaline Phosphatase is the enzyme activity which hydrolyzes 1 mol of 4-nitrophenyl phosphatase in one minute at +37 °C under assay conditions.

Volume Activity: 1 U/μl

Packaging

1 kit containing 2 components

Kit Components Only

Product No.
Description

  • rAPid Alkaline Phosphatase Buffer 10x concentrated

  • rAPid Alkaline Phosphatase 1 U/μl

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel Wells et al.
Molecular breeding : new strategies in plant improvement, 33, 349-362 (2014-02-04)
Many important plant species have polyploidy in their recent ancestry, complicating inferences about the genetic basis of trait variation. Although the principal locus controlling the proportion of polyunsaturated fatty acids (PUFAs) in seeds of Arabidopsis thaliana is known (fatty acid
Shiwen Jiang et al.
Retrovirology, 6, 73-73 (2009-08-12)
Human T-cell leukemia virus type 1 (HTLV-1) causes adult T -cell leukemia (ATL) but the expression of HTLV-1 is strongly suppressed in the peripheral blood of infected people. However, such suppression, which may explain the long latency in the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service