Skip to Content
Merck

LEU-RO

Roche

Leupeptin

powder, from synthetic, =96.5%

Synonym(s):

Leupeptin hemisulfate salt, Acetyl-Leu-Leu-Arg-al, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H38N6O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
475.59
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

description

Ac-Leu-Leu-argininal x 1/2 H2SO4, synthetic

assay

96.5%

form

powder

mol wt

Mr 475.6

packaging

pkg of 100 mg (11529048001), pkg of 25 mg (11017128001), pkg of 5 mg (11017101001), pkg of 50 mg (11034626001)

manufacturer/tradename

Roche

solubility

H2O: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O

InChI

1S/2C20H38N6O4.H2O4S/c2*1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;1-5(2,3)4/h2*11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);(H2,1,2,3,4)/t2*15-,16-,17-;/m00./s1

InChI key

CIPMKIHUGVGQTG-VFFZMTJFSA-N

Application

Leupeptin as well as other protease inhibitors like antipain, chymostatin, pepstatin, and phosphoramidon are useful for the protection of proteins during their isolation from tissues or membranes. Leupeptin can be removed from the reaction by dialysis.
Note: To check other protease inhibitors, try our Protease Inhibitor Set including Antipain Dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pefabloc SC, Pepstatin, and Phosphoramidon.

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM.
Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)
Leupeptin inhibits serine and thiol proteases such as trypsin, plasmin, proteinase K, kallikrein, papain, thrombin, and cathepsin A and B.
Not affected are α-, β-, γ- and δ- chymotrypsin, pepsin, cathepsin D, elastase, renin, and thermolysin.

Features and Benefits

Contents
Synthetic, white powder

Preparation Note

Working concentration: 0.5 to 5 μg/ml (1 to 20 μM)

Comparison of working concentrations of pefabloc with leupeptin and PMSF is in files.

Working solution: Solvent is recommended in distilled water.
Highly soluble in water (1 mg/ml), methanol, ethanol, acetic acid, dimethyl formamide and dimethyl sulfoxide.
Poorly soluble in acetone, chloroform, ethyl ether and n-hexane.

Storage conditions (working solution): -15 to -25 °C
In aqueous solution leupeptin is stable for 1 month at 2 to 8 °C or for at least 6 months at - 15 to -25 °C, stored under nitrogen. For best results, freeze the dissolved inhibitor in aliquots and avoid repeated thawing.
Highly soluble in water (1 mg/ml). Stable for at least one week at 2 to 8 °C and 6 months frozen in aliquots at -15 to -25 °C.

Analysis Note

Function test: Performance controlled with trypsin.
Leupeptin gives multiple peaks on HPLC due to equilibria among three forms in solution. Purity determined using three main peaks. Majority of contaminating peptide is racemized leupeptin.

Other Notes

For life science research only. Not for use in diagnostic procedures.


Still not finding the right product?

Explore all of our products under Leupeptin


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
1101710100104061838694904
1101712800104061838694911
1152904800104061838703217
1103462600104061838695949