Select a Size
About This Item
Skip To
grade
Molecular Biology
assay
99.8% (sum of 4 peaks, HPLC)
form
solution
usage
sufficient for 2,500 reactions (04638956001), sufficient for 5,000 reactions (11814362001), sufficient for 500 reactions (11581295001)
packaging
pkg of 10 × 200 μL (11814362001 [10mM, each]), pkg of 200 μL (11581295001 [10 mM, each]), pkg of 5 × 200 μL (04638956001 [10 mM, each])
manufacturer/tradename
Roche
concentration
10 mM
impurities
DNase, none detected, RNase, none detected
color
colorless
solubility
water: miscible
storage temp.
−20°C
1 of 4
This Item | 11888412001 | 72004 | 11585550910 |
|---|---|---|---|
| grade Molecular Biology | grade - | grade - | grade - |
| manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename - | manufacturer/tradename Roche |
| Quality Level 100 | Quality Level 100 | Quality Level 300 | Quality Level 100 |
| assay 99.8% (sum of 4 peaks, HPLC) | assay - | assay - | assay - |
| form solution | form solution | form liquid | form solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
General description
Application
Features and Benefits
Improve yield and performance
- Choose a convenient ready-to-use mix of all 4 nucleotides.
- Achieve the highest PCR and RT-PCR sensitivity.
- Insist on highest purity (>99%) nucleotides.
Packaging
Preparation Note
Working solution: If required, the solutions can be diluted with distilled autoclaved water.
Storage conditions (working solution): It will withstand 50 freeze/thaw cycles.
Analysis Note
Function Testing in RT-PCR
Absence of Contaminating Deoxyribonucleases/Nicking Activities according to the current Quality Control procedures.
Absence of Contaminating Ribonucleases according to the current Quality Control procedures.
Other Notes
Kit Components Only
- dATP, PCR Grade, sodium salt 10 mM
- dCTP, PCR Grade, sodium salt 10 mM
- dGTP, PCR Grade, sodium salt 10 mM
- dTTP, PCR Grade, sodium salt 10 mM
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


