Skip to Content
Merck

Skip To

DNALIG-RO

Roche

T4 DNA Ligase

solution, from Escherichia coli(NM 989), suitable for activity assay

Synonym(s):

dna ligase, t4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Specific activity:
~3000 units/mg protein
Biological source:
Escherichia coli (NM 989)
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli (NM 989)

recombinant

expressed in E. coli

description

recombinant form of the enzyme from T4 phage

form

solution

specific activity

~3000 units/mg protein

packaging

pkg of 100 U (10481220001 [1 U/μl]), pkg of 500 U (10716359001 [1 U/μl]), pkg of 500 U (10799009001 [5 U/μl])

manufacturer/tradename

Roche

parameter

4-22 °C optimum reaction temp.

technique(s)

activity assay: suitable

optimum pH

7.2-7.8

storage temp.

−20°C

General description

T4 DNA Ligase catalyzes the formation of phosphodiester bonds between neighboring 3′-hydroxyl- and 5′-phosphate ends in double-stranded DNA. Single-stranded nicks in double-stranded DNA are also closed by T4 DNA Ligase.

Contents
T4 DNA Ligase, supplied with 10x concentrated ligation buffer that includes ATP.

Application

Ligation of sticky- and blunt-ended DNA fragments.[1][2][3]

Biochem/physiol Actions

Heat inactivation: The enzyme can be inactivated by heating (65 °C, 10 minutes).

Preparation Note

Activator: Mg2+ (10mM), DTT
Working solution: For dilution of the enzyme Roche recommends using a buffer containing the components of the storage buffer: 20 mM Tris-HCl, 60 mM KCl, 1 mM EDTA, 5 mM dithoerythritol, 50% glycerol (v/v), pH 7.5 (4 °C).

Analysis Note

Quality

Tested for the absence of deoxyribonucleases and exonucleases, according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.
One unit of T4 DNA ligase is the amount of enzyme activity that converts 1 nmol 32P from pyrophosphate into Norit-absorbable material in 20 minutes at +37 °C. One unit corresponds to 0.2 U determined in the exonuclease III resistance assay. Dilution buffer: 50 mM Tris-HCl, 10 mM dithioerythritol, bovine serum albumin, 500 μg/ml, pH 7.6 (+25 °C).

Volume Activity: 1 x 103 U/ml; 5 x 103 U/ml

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GE27-0361-01D2886RDLIG-RO
biological source

Escherichia coli (NM 989)

biological source

-

biological source

-

biological source

-

technique(s)

activity assay: suitable

technique(s)

-

technique(s)

-

technique(s)

-

specific activity

~3000 units/mg protein

specific activity

-

specific activity

4,000 U/mL

specific activity

-

form

solution

form

-

form

buffered aqueous glycerol solution

form

-

recombinant

expressed in E. coli

recombinant

-

recombinant

-

recombinant

-

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library