Skip to Content
Merck

CRPHO-RO

Roche

Creatine phosphate

crystalline, >97%

Synonym(s):

Sodium creatine phosphate dibasic tetrahydrate, Creatine phosphate disodium salt tetrahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Na2O3PNHC(=NH)N(CH3)CH2COOH · 4H2O
CAS Number:
Molecular Weight:
327.14
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>97%

form

crystalline

mol wt

327.14

packaging

pkg of 10 g (10621722001), pkg of 5 g (10621714001)

manufacturer/tradename

Roche

SMILES string

O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(=N)NP([O-])([O-])=O

InChI

1S/C4H10N3O5P.2Na.4H2O/c1-7(2-3(8)9)4(5)6-13(10,11)12;;;;;;/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);;;4*1H2/q;2*+1;;;;/p-2

InChI key

HUWYWJSJJDCZRQ-UHFFFAOYSA-L

General description

Creatine kinase is responsible for the production of ATP (adenosine triphosphate) by transferring phosphate from creatine phosphate to ADP (adenosine diphosphate). Creatine phosphate is provided as a disodium salt.

Application

Creatine phosphate has been used in the energy regeneration mix for sperm, spermatids and egg extracts.

Analysis Note

Purity: >97% creatine-P-Na2 x 4 H2O

Other Notes

For life science research only. Not for use in diagnostic procedures.


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
1062171400104061838673824
1062172200104061838673862