11417959001
Roche
SuRE/Cut™ Buffer A
pH 8.1-8.3 (37 °C), suitable for molecular cloning
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105600
form
solution
packaging
pkg of 5 × 1 mL
manufacturer/tradename
Roche
technique(s)
molecular cloning: suitable
pH
8.1-8.3 (37 °C)
storage temp.
−20°C
General description
The SuRE/Cut Buffer System consists of five optimized incubation buffers, 10x concentrated, for DNA restriction digest. Activity of all restriction enzymes has been determined in each buffer to select 100% activity or to calculate activity in double-digests.
Application
SuRE/Cut™ Buffer A is suitable for DNA restriction digest.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Legal Information
SuRE/Cut is a trademark of Roche
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
The term “Restriction enzyme” originated from the studies of Enterobacteria phage λ (lambda phage) in the laboratories of Werner Arber and Matthew Meselson.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service