Skip to Content
Merck
All Photos(1)

Key Documents

11388908910

Roche

Biotin-16-UTP

pkg of 25 μL (250 nmol; 10mM)

Synonym(s):

Biotin-16-UTP, biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

Assay

97.7% (HPLC)

Quality Level

form

solution

mol wt

Mr 987.5 (biotin-16-UTP-Li4)

packaging

pkg of 25 μL (250 nmol; 10mM)

manufacturer/tradename

Roche

color

colorless

solubility

water: miscible

shipped in

dry ice

storage temp.

−20°C

General description

For biotin-16-UTP, biotin is bound to uridine triphosphate via an amide linkage. Biotin-16-UTP is a substrate for SP6, T3, and T7 RNA polymerase. It can replace UTP in the in vitro transcription reaction for RNA labeling. Linearized template DNA with T7, SP6, or T3 promoter is in vitro-transcribed with the corresponding RNA polymerases using ATP, GTP, CTP, UTP, and Biotin-16-UTP, respectively.

Application

Biotin-labeled RNA can be used as a hybridization probe for:
  • Northern blots
  • Southern blots
  • Plaque or colony lifts
  • RNase protection experiments
  • In situ hybridization
  • Microarray hybridization
The labeled RNA can be subsequently detected with a fluorescent streptavidin conjugate or by enzyme linked immuno sorbent assay (ELISA) using a Streptavidin-AP conjugate.

Quality

Typical analysis: At least 85% Biotin-16-UTP (HPLC, area%).

Physical form

Biotin-16-UTP, tetralithium salt, 10mM solution

Other Notes

For life science research only. Not for use in diagnostic procedures.

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damian LaGamba et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 234(1), 132-142 (2005-07-13)
One of the most fundamental biological processes in development, as well as a primary mechanism for tumor metastasis, is epithelial-mesenchymal transformation (EMT). To gain a greater understanding of this transition, we have obtained a genomic profile of the critical stages
Chenying Li et al.
Haematologica, 105(1), 148-160 (2019-04-13)
Homoharringtonine, a plant alkaloid, has been reported to suppress protein synthesis and has been approved by the US Food and Drug Administration for the treatment of chronic myeloid leukemia. Here we show that in acute myeloid leukemia (AML), homoharringtonine potently
Subramaniam Jayanthi et al.
PloS one, 4(6), e6092-e6092 (2009-07-01)
Methamphetamine (METH) is an illicit toxic psychostimulant which is widely abused. Its toxic effects depend on the release of excessive levels of dopamine (DA) that activates striatal DA receptors. Inhibition of DA-mediated neurotransmission by the DA D1 receptor antagonist, SCH23390
Aikaterini Tsaousi et al.
Circulation research, 108(4), 427-436 (2011-01-05)
Vascular smooth muscle cell (VSMC) proliferation causes intimal thickening in atherosclerosis and restenosis. Previously, we demonstrated that Wnt/β-catenin signaling upregulates VSMC proliferation in vitro. We examined this pathway in vivo and investigated the involvement of specific Wnt proteins in VSMC
Julie Bergalet et al.
Cell reports, 30(10), 3339-3352 (2020-03-12)
Overlapping genes are prevalent in most genomes, but the extent to which this organization influences regulatory events operating at the post-transcriptional level remains unclear. Studying the cen and ik2 genes of Drosophila melanogaster, which are convergently transcribed as cis-natural antisense

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service