Skip to Content
Merck
All Photos(1)

Key Documents

10884308001

Roche

CPRG

Chlorophenol red-β-D-galactopyranoside

Synonym(s):

Chlorophenol Red-β-D-galactopyranoside, chlorophenol red-β-d-galactopyranoside, CPRG, Chlorophenol Red-β-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22Cl2O10S
CAS Number:
Molecular Weight:
585.41
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Quality Level

Assay

96% (HPLC)

form

powder

packaging

pkg of 250 mg

manufacturer/tradename

Roche

storage condition

protect from light

pH range

6-8

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2Cl)C(=C3/C=CC(=O)C(Cl)=C3)\c4ccccc4S(O)(=O)=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C25H22Cl2O10S/c26-15-9-12(5-7-17(15)29)21(14-3-1-2-4-20(14)38(33,34)35)13-6-8-18(16(27)10-13)36-25-24(32)23(31)22(30)19(11-28)37-25/h1-10,19,22-25,28,30-32H,11H2,(H,33,34,35)/b21-12-/t19-,22+,23+,24-,25-/m1/s1

InChI key

YOUWVNCQJOMMEU-AETRGLENSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chlorophenol red-β-D-galactopyranoside (CPRG) is a long-wavelength dye which is commonly used for colorimetric assays. The reaction product has a dark red color and is soluble in water. It can be evaluated photometrically at 574 or 578 nm.
The chemical formula on the label represents the monosodium salt.

Application

CPRG has been used as a substrate for β-galactosidase assay.
Colorimetric determination of β-Gal activity in cell extracts and ELISA.

Biochem/physiol Actions

Chlorophenol red-β-D-galactopyranoside (CPRG) might possess oestrogenic activity.

Quality

Contaminant: <0.1% chlorophenol red

Sequence

CPRG is an approximately 1:1 mixture of the isomers.

Preparation Note

Working concentration: 5 mmol/l
Storage conditions (working solution): Reconstituted solution is stable for one day at 2 to 8 °C

Storage and Stability

Store powder dry at -15 to -25 °C; stock solutions at 2–8 °C protected from light.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias A Fügi et al.
Journal of lipid research, 55(5), 929-938 (2014-03-15)
Sterols are an essential class of lipids in eukaryotes, where they serve as structural components of membranes and play important roles as signaling molecules. Sterols are also of high pharmacological significance: cholesterol-lowering drugs are blockbusters in human health, and inhibitors
Els Vanderperren Wim Demaré Ronny Blust Kris Cooreman Peter Bossier
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 6(5), 375-380 (2001-01-01)
The finding that a variety of chemicals display oestrogenic activity has resulted in the development of in vitro and in vivo assays to assess oestrogenic activity. One such assay, the yeast oestrogen assay (YES) makes use of recombinant yeast cells
Mallika Ghosh et al.
Bio-protocol, 2(24), e305-e305 (2012-12-20)
Antigen cross presentation is important for effective immune responses to tumors and viral infections. Dendritic cells are professional antigen presenting cells and are unique in their ability to cross-present exogenous antigens on MHC class I molecules and activate antigen specific
Evan Amalfitano et al.
Nature communications, 12(1), 724-724 (2021-02-03)
Recent advances in cell-free synthetic biology have given rise to gene circuit-based sensors with the potential to provide decentralized and low-cost molecular diagnostics. However, it remains a challenge to deliver this sensing capacity into the hands of users in a
Andrés Alloatti et al.
Bio-protocol, 6(22) (2017-02-28)
Antigen presentation by MHC class I molecules, also referred to as cross-presentation, elicits cytotoxic immune responses. In particular, dendritic cells (DC) are the most proficient cross-presenting cells, since they have developed unique means to control phagocytic and degradative pathways. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service