Skip to Content
Merck

TR-1005

Cloning Cylinder, 10 mm x 10 mm diameter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32011202
Manufacturer/tradename:
Specialty Media
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Specialty Media

Quality Segment

input

sample type: mouse embryonic stem cell(s)

shipped in

ambient

storage temp.

15-25°C

General description

Store at ambient temp.

Application

Research Category
Cell Culture

Packaging

10 sterile cylinders/60mm dish

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library