Skip to Content
Merck
All Photos(1)

Key Documents

ST1202

Sigma-Aldrich

Anti-PGC-1α Mouse mAb (4C1.3)

liquid, clone 4C1.3, Calbiochem®

Synonym(s):

Anti-Peroxisome Proliferator-Activated Receptor- γ Coactivator 1 α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4C1.3, monoclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

General description

PGC-1α is a co-activator of PPARα, PPARγ, and other transcription factors and regulates the transcriptonal program of adaptive thermogenesis in brown adipose tissue, hepatic/renal gluconeogenesis, and muscle fiber type switching. The full-length protein is 113 kDa and is induced in brown adipose tissue by cold exposure, in liver and kidney by fasting, and in skeletal muscle by exercise. Alternative splicing of the full-length gene produces a 270 aa N-terminal splice variant that migrates at ~38 kDa and is induced by the same physiological signals that induce expression of the full-length protein. The N-terminal PGC-1α splice variant is also subject to post translational modifications that alter its migration and apparent molecular weight.
Protein G purified mouse monclonal antibody. Recognizes the endogenous forms of PGC-1α, which includes the ~113 kDa PGC-1α protein and the ~38 kDa splice variant.
Recognizes the endogenous forms of PGC-1α, which includes the ~113 kDa PGC-1α protein and the ~38 kDa splice variant, in brown adipose tissue, liver, and kidney.
This Anti-PGC-1α Mouse mAb (4C1.3) is validated for use in Immunoblotting, Immunocytochemistry, Immunoprecipitation, Paraffin Sections for the detection of PGC-1α.

Immunogen

Mouse
a recombinant protein consisting of amino acids 1-120 of mouse PGC-1α

Application

Immunoblotting (1 µg/ml)

Immunocytochemistry (5-10 µg/ml)

Immunoprecipitation (10 µg/ml)

Paraffin Sections (whole mount) (see application references)

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Physical form

One vial of 100 µg antibody in 50 mM PBS, see vial for lot-specific concentration. One vial of NT-PGC-1α Positive Control, supplied as 25 µl whole cell extract in RIPA buffer containing 10 mM &beta-mercaptoethanol and 1% SDS; load 10 µl per lane; add sample buffer prior to SDS-PAGE loading.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Endogenous PGC-1α is found primarily in the nucleus and NT-PGC-1α is found in both cytosolic and nuclear fractions. Antibody should be titrated for optimal results in individual systems.
Lai, L., et al. 2008. Genes Dev. 14, 1948.
Rodgers, J.T., et al. 2008. FEBS Lett. 582, 46.
Mazzucotelli, A., et al. 2007. Diabetes 10, 2467.
Nemoto, S., et al. 2005. J. Biol. Chem. 16, 16456.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Horii et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 312(4), R520-R528 (2017-01-27)
The purpose of this study was to investigate the effect of chronic chlorella intake alone or in combination with high-intensity intermittent exercise (HIIE) training on exercise performance and muscle glycolytic and oxidative metabolism in rats. Forty male Sprague-Dawley rats were
Amy E Mendham et al.
Journal of applied physiology (Bethesda, Md. : 1985), 121(6), 1326-1334 (2016-10-16)
This study assessed the mitochondrial related signaling responses to a single bout of noncontact, modified football (touch rugby), played as small-sided games (SSG), or cycling (CYC) exercise in sedentary, obese, middle-aged men. In a randomized, crossover design, nine middle-aged, sedentary
Chi Luo et al.
The Journal of clinical investigation, 130(2), 853-862 (2020-01-14)
Oncogene-targeted and immune checkpoint therapies have revolutionized the clinical management of malignant melanoma and now offer hope to patients with advanced disease. Intimately connected to patients' overall clinical risk is whether the initial primary melanoma lesion will metastasize and cause
James R Broatch et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 313(4), R372-R384 (2017-07-07)
We investigated the underlying molecular mechanisms by which postexercise cold-water immersion (CWI) may alter key markers of mitochondrial biogenesis following both a single session and 6 wk of sprint interval training (SIT). Nineteen men performed a single SIT session, followed
Dean-Chuan Wang et al.
International journal of molecular sciences, 21(11) (2020-06-04)
Early-life exposure to di-(2-ethylhexyl)-phthalate (DEHP) has been suggested to relate to hyperactivity, lack of attention, and working memory deficits in school-age children. Brain-derived neurotrophic factor (BDNF) and endocannabinoids are induced by aerobic exercises to provide beneficial effects on brain functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service