Skip to Content
Merck

SM-2001

Trypsin

The Trypsin 0.25%, In Hank′s Balanced Salt Solution, without Ca 2+ & Mg 2+

Synonym(s):

Proteolytic enzyme, Trypsin enzyme, Trypsin solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160801
NACRES:
NA.75
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Trypsin 0.25%, In Hank′s Balanced Salt Solution, without Ca2+ and Mg2+, The Trypsin 0.25%, In Hank′s Balanced Salt Solution, without Ca 2+ & Mg 2+ is available in a 100 mL format.

Quality Segment

sterility

non-sterile

form

liquid

manufacturer/tradename

Specialty Media

input

sample type: mouse embryonic stem cell(s)
sample type cardiac stem cell(s)
sample type hematopoietic stem cell(s)
sample type pancreatic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells

Preparation Note

Store at -20°C for 18 months.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library