Skip to Content
Merck

Skip To

SCM044

FibroGRO® Xeno-Free Human Fibroblast Expansion Medium

The FibroGRO Xeno-Free Human Fibroblast Expansion Medium is a new variation of our FibroGRO media that is optimized for the culture of fibroblasts without the use of animal-derived components.

Synonym(s):

Primary Cell Culture Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Chemicon®, FibroGRO®, Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-10 to -25°C

General description

FibroGRO Xeno-Free Human Fibroblast Expansion Medium is a new variation of our FibroGRO cell culture media that is optimized for the culture of fibroblasts without the use of animal-derived components. EMD Millipore’s FibroGRO cell system is perfect for establishing Xeno-Free human feeder layers for induced pluripotent, embryonic, and general stem cell culture. It can also be used to establish a model to study wound healing, toxicology and basic cell biology. FibroGRO Xeno Free supports the growth of these cells in a 2% human serum environment at rates equal to or greater than comparable media supplemented with 2-10% FBS. Our FibroGRO Xeno-Free media does not contain any antimicrobials, phenol red, or any other components that can cause cell stress and “masking effects” that may influence experimental results.

Application

Research Category
Stem Cell Research

Physical form

FibroGROTM Xeno-Free Human Fibroblast Expansion medium is sold as a complete frozen medium and contains all the necessary growth factors and supplements to support fibroblast growth.

Preparation Note

Frozen medium should be stored at -20ºC until expiration date.
Thawed medium can be stored at 2 to 8ºC for up to 1 month.
Multiple freeze/thaw cycles are not recommended

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
FIBROGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCM045SCM037C-23120
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

manufacturer/tradename

Chemicon®, Millipore

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®, Millipore

manufacturer/tradename

-


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters