Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Skip To

SCM005

ReNcell NSC Maintenance Media

ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free neural stem cell medium that has been optimized for the growth & in vitro differentiation of ReNcell immortalized human neural progenitor cells.

Synonym(s):

ReNcell Neural Stem Cell Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

dry ice

storage temp.

-10 to -25°C

General description

ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells. When used in conjunction with FGF and EGF, the maintenance medium will allow for the proliferation of ReNcell immortalized VM and CX neural stem cells. Withdrawal of the growth factors from ReNcell NSC Maintenance Medium will result in the spontaneous differentiation of ReNcell immortalized neural progenitor cells

Physical form

ReNcell NSC Maintenance Medium contains DMEM/F12, HEPES, L-glutamine, human serum albumin, human transferrin, putrescine dihydrochloride, human recombinant insulin, L-thyroxine, Tri-iodo-thyronine, progesterone, sodium selenite, heparin, corticosterone. The media does not contain antibiotics.

Preparation Note

SCM005, ReNcell Neural Stem Cell Maintenance Medium should be stored at -20°C until ready to use. Upon thawing, the maintenance medium should be stored at 2-8°C and given a 1-month expiration dating.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCM003SCM004SCM101
description

ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free neural stem cell medium that has been optimized for the growth & in vitro differentiation of ReNcell immortalized human neural progenitor cells.

description

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth & in vitro differentiation of neural stem cells derived from rodents.

description

The ENStem-A Neural Expansion Medium is a defined serum-free formulation that has been optimized for the culture & expansion of ENStem-A Human Neural Progenitor Cells.

description

The Cardiac Stem Cell Maintenance Medium is available in a 500 mL format & has been optimized & validated for cell culture.

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters