Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
NACRES:
NA.28
eCl@ss:
32011203
Technique(s):
cell culture | mammalian: suitable
description
Cell line type: Cancer Cells
technique(s)
cell culture | mammalian: suitable
General description
Ovarian carcinoma is a prevalent worldwide disease, with over 200,000 new cases diagnosed per year (1). The first-line chemotherapy for newly-diagnosed ovarian carcinoma is cisplatin, but despite high initial efficacy, the most common types of ovarian carcinomas frequently relapse into platinum-resistant forms and many demonstrate multi-drug resistance (1). The availability of cellular models that recapitulate both resistant and susceptible forms of the disease are essential to understanding drug resistance and advancing new options for treatment.
OVCAR-4 is a high-grade serous ovarian adenocarcinoma cell line established from a patient refractory to cisplatin, and is resistant to multiple chemotherapeutic agents (2). OVCAR-4 cells are characterized by migration/invasion ability and are tumorigenic in nude mice (3). The OVCAR-4 cell line is homozygous for the Leu130Val mutation in TP53 (4) and expresses high levels of the cancer marker integrin αvβ3 (5). OVCAR-4 cells are recommended to be passaged at relatively high density (at least 13,000 cells/cm2). OVCAR-4 is one of the most representative cell lines for high-grade serous ovarian carcinoma and a highly relevant model for multidrug-resistant ovarian cancer.
Source:
OVCAR-4 was established from ascites from a 42-year-old patient with an ovarian tumor refractory to cisplatin treatment (2).
References:
1. Nat Rev Disease Primers 2016; 2: 16061.
2. Cancer Res 1997; 57: 850—856.
3. Gynecol Oncol 2015;138(2): 372-377.
4. Mol Cancer Ther 2006; 5(11): 2606-2612.
5. Bioorganic & Medicinal Chem 2018; 26: 2085-2091.
OVCAR-4 is a high-grade serous ovarian adenocarcinoma cell line established from a patient refractory to cisplatin, and is resistant to multiple chemotherapeutic agents (2). OVCAR-4 cells are characterized by migration/invasion ability and are tumorigenic in nude mice (3). The OVCAR-4 cell line is homozygous for the Leu130Val mutation in TP53 (4) and expresses high levels of the cancer marker integrin αvβ3 (5). OVCAR-4 cells are recommended to be passaged at relatively high density (at least 13,000 cells/cm2). OVCAR-4 is one of the most representative cell lines for high-grade serous ovarian carcinoma and a highly relevant model for multidrug-resistant ovarian cancer.
Source:
OVCAR-4 was established from ascites from a 42-year-old patient with an ovarian tumor refractory to cisplatin treatment (2).
References:
1. Nat Rev Disease Primers 2016; 2: 16061.
2. Cancer Res 1997; 57: 850—856.
3. Gynecol Oncol 2015;138(2): 372-377.
4. Mol Cancer Ther 2006; 5(11): 2606-2612.
5. Bioorganic & Medicinal Chem 2018; 26: 2085-2091.
Application
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.
Preparation Note
Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Analysis Note
- Each vial contains >=1X106 viable cells.
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are negative for mycoplasma contamination.
- Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.