Skip to Content
Merck

SCC237

YUMM4.1 Mouse Melanoma Cell Line

Mouse

Synonym(s):

Yale University Mouse Melanoma 2.1 cell line, Yale University Mouse Melanoma cell line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
mouse
Manufacturer/tradename:
Millipore
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

YUMM4.1 Mouse Melanoma Cell Line,

biological source

mouse

Quality Segment

packaging

vial of ≥1X10⁶ cells/vial  vials

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

The YUMM4.1 mouse melanoma cell line was derived from a 4-hydroxytamoxifen-induced melanoma tumor in a female C57/Bl/6 mouse into which mutations from the Braf/Pten genetically-engineered mouse model had been introduced via backcrossing. The YUMM4.1 cell line is homozygous negative for Pten and Cdkn2.1?
The promise of immune-based therapies in cancer and increasingly successful application of these approaches have emphasized the necessity of immune-competent models to evaluate immunological responses to cancer cells. Immunocompetent genetically-engineered mouse models with distinct genetic drivers of melanoma are essential for identifying potential immunotherapies, but are limited by the need to maintain colonies of multiple genotypes necessary to generate mouse models with appropriate genetic backgrounds.YUMM4.1 cells are syngenic with the immunocompetent C57BL/6 mouse background and retain genetic markers of the Braf/Pten mouse model. Although mutations in Braf V600 are prevalent in melanomas and are targets of specific drugs, first-line treatment of non-Braf mutant melanoma consists of immune checkpoint inhibitors, to which roughly half of patients do not respond. The YUMM4.1 cell line is homozygous negative for Pten and Cdkn2 but has no activating Braf mutations, making this cellular model ideal for investigating the biology of non-Braf-mutant melanomas and potential therapeutics for this difficult subset of cancer.​Mutation: Pten−/− Cdkn2−/−​ . Female.References:1. Meeth K et al. (2016) The YUMM lines: a series of congenic mouse melanoma cell lines with defined genetic alterations. Pigment Cell Melanoma Res 29(5): 590-597.2. Dankort D et al. (2009) Braf(V600E) cooperates with Pten loss to induce metastatic melanoma. Nat Genet. 41(5): 544-552.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.

Features and Benefits

The YUMM cell lines recapitulate genetic drivers found in many human melanomas. YUMM4.1 is homozygous negative for Pten and Cdkn2+, and has no activating Braf mutations.

Preparation Note

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library