Skip to Content
Merck
All Photos(2)

Key Documents

SCC180

Sigma-Aldrich

OECM-1 Human Oral Squamous Carcinoma Cell Line

Human

Synonym(s):

OECM1, OEC-M1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

OECM-1 Human Oral Squamous Carcinoma Cell Line, OECM-1 human oral squamous carcinoma cell line is suitable for studies of cancer cell signaling, epithelial-mesenchymal transition (EMT), metastasis, invasion, and cancer cell stemness.

biological source

human

technique(s)

cell culture | mammalian: suitable

General description

Squamous cell carcinoma (SCC) is a common cancer type, developing in epithelial cells. Nearly all cancers of the oral cavity are squamous cell carcinomas, and the high recurrence rate among oral SCCs make these challenging targets for therapies. Established models of this cancer type continue to yield significant insights into the properties and potential treatments of head and neck cancers .
The OECM-1 human oral cavity squamous cell carcinoma cell line is a well-established model for squamous cell carcinoma. The OECM-1 cell line harbors a missense mutation in the p53 tumor suppressor , displays low EGFR expression , and is tumorigenic in nude mice . OECM-1 cells proliferate with a doubling time of 30-38 hours and are capable of anchorage-independent growth, forming spheroid colonies . OECM-1 cells have been intensively characterized in the literature for morphology, biomarker expression and drug response .
Source:
The OECM-1 human oral cavity squamous cell carcinoma cell line was derived from surgical resection of a primary tumor of a Taiwanese male patient .

Cell Line Description

Cancer Cells

Application

OECM-1 human oral squamous carcinoma cell line is suitable for studies of cancer cell signaling, epithelial-mesenchymal transition (EMT), metastasis, invasion, and cancer cell stemness.
Research Category
Cancer
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X10⁶ viable cells.
• Cells are tested negative for HPV-16, HPV-18, Hepatitis A, B, C, and HIV-1 & 2 viruses by PCR.
• Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

OECM-1 human oral cavity squamous carcinoma cell line should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Wen Chang et al.
Cell death & disease, 9(2), 194-194 (2018-02-09)
Cancer-initiating cells (CICs) are responsible for tumor initiation, progression, and therapeutic resistance; moreover, redox homeostasis is important in regulating cancer stemness. Previously, we have identified that cancer cells containing low intracellular reactive oxygen species levels (ROSLow cells) display enhanced features
Wei Yan et al.
Frontiers in cell and developmental biology, 9, 616306-616306 (2021-04-09)
Oral squamous cell carcinoma (OSCC), accounting for two-thirds of head and neck cancer, is characterized by poor prognosis and a high rate of mortality. Exosomes have emerged as potential molecule-shuttle in intercellular communication, thereby regulating the physiological processes of recipient
Shu-Chun Lin et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 33(2), 79-86 (2004-01-15)
Cell lines derived from oral squamous cell carcinoma (OSCC) exposed to variable etiological factors can bestow advantages in understanding the molecular and cellular alterations pertaining to environmental impacts. Most OSCC cell lines have been established from smoker patients or areca
Benoite Méry et al.
Oral oncology, 65, 51-56 (2017-01-23)
Head and neck cancer remains a significant public health concern. About 60% of patients die within 5years due to local recurrence. Head and neck squamous cell carcinoma (HNSCC) cell lines are important preclinical models in the search for new therapies
Guan-Cheng Huang et al.
Japanese journal of clinical oncology, 34(9), 499-504 (2004-10-07)
Platinum, 5-fluorouracil (5-FU) and taxanes are commonly used in chemotherapeutic modalities of various carcinomas. However, taxanes are rarely used in patients suffering from head and neck squamous cell carcinoma (HNSCC) in Taiwan. The purpose of this study was to assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service