Skip to Content
Merck
All Photos(1)

Key Documents

S7101

Sigma-Aldrich

ApopTag Plus Peroxidase In Situ Apoptosis Kit

The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling & detecting DNA strand breaks by the indirect TUNEL method.

Synonym(s):

In Situ Apoptosis Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84
Pricing and availability is not currently available.

Quality Level

manufacturer/tradename

ApopTag
Chemicon®

detection method

colorimetric

shipped in

dry ice

General description

The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling and detecting DNA strand breaks by the indirect TUNEL method. The kit provides sufficient reagents for immunoperoxidase staining of 40 samples, and includes ApopTag(R) Positive control Slides and DAB buffer and substrate. Results are visualized using brightfield microscopy.

The ApopTag Peroxidase Kits have been qualified for use in histochemical and cytochemical staining of the following specimens: formalin-fixed, paraffin-embedded tissues, cryostat sections, cell suspensions, cytospins, and cell cultures. Whole mount-methods have been developed (34, 45). (See datasheet Sec. V. for References).

The staining specificity of the ApopTag Peroxidase Kits has been demonstrated by Chemicon and many other laboratories. Chemicon has tested many types of model cell and tissue systems, including: (a) human prostate, thymus, and large intestine (in-house data); (b) rat ventral prostate post-castration (21), (c) rat thymus lymphocytes treated in vitro with dexamethasone (3, 13), (d) 14-day mouse embryo limbs (1) and (e) rat mammary gland in regression after weaning (36). In the thymocyte and prostate models, agarose gel electrophoresis was used to assess the amount of DNA laddering, which peaked coincidentally with the maximum percentage of stained cells. Numerous journal publications from laboratories worldwide have established the usefulness of ApopTag Kits. (See datasheet Sec. V. References, Publications Citing ApopTag Kits).

Application

INTRODUCTION

ApopTag In Situ Apoptosis Detection Kits label apoptotic cells in research samples by modifying DNA fragments utilizing terminal deoxynucleotidyl transferase (TdT) for detection of apoptotic cells by specific staining.

This manual contains information and protocols for the ApopTag Plus In Situ Apoptosis Detection Kit.

Principles of the Procedure

The reagents provided in ApopTag Peroxidase Kits are designed to label the free 3′OH DNA termini in situ with chemically labeled and unlabeled nucleotides. The nucleotides contained in the Reaction Buffer are enzymatically added to the DNA by terminal deoxynucleotidyl transferase (TdT) (13, 31). TdT catalyzes a template-independent addition of nucleotide triphosphates to the 3′-OH ends of double-stranded or single-stranded DNA. The incorporated nucleotides form an oligomer composed of digoxigenin-conjugated nucleotide and unlabeled nucleotide in a random sequence. The ratio of labeled to unlabeled nucleotide in ApopTag Peroxidase Kits is optimized to promote anti-digoxigenin antibody binding. The exact length of the oligomer added has not been measured.

DNA fragments which have been labeled with the digoxigenin-nucleotide are then allowed to bind an anti-digoxigenin antibody that is conjugated to a peroxidase reporter molecule (Figure 1A). The bound peroxidase antibody conjugate enzymatically generates a permanent, intense, localized stain from chromogenic substrates, providing sensitive detection in immunohistochemistry or immunocytochemistry (i.e. on tissue or cells). This mixed molecular biological-histochemical systems allows for sensitive and specific staining of very high concentrations of 3′-OH ends that are localized in apoptotic bodies.



The ApopTag system differs significantly from previously described in situ labeling techniques for apoptosis (13, 16, 38, 46), in which avidin binding to cellular biotin can be a source of error. The digoxigenin/anti-digoxigenin system has been found to be equally sensitive to avidin/biotin systems (22). The sole natural source of digoxigenin is the digitalis plant. Immunochemically-similar ligands for binding of the anti-digoxigenin antibody are generally insignificant in animal tissues, ensuring low background staining. Affinity purified sheep polyclonal antibody is the specific anti-digoxigenin reagent used in ApopTag Kits. This antibody exhibits <1% cross-reactivity with the major vertebrate steroids. In addition, the Fc portion of this antibody has been removed by proteolytic digestion to eliminate any non-specific adsorption to cellular Fc receptors.

Results using ApopTag Kits have been widely published (see Sec. V. References, Publications Citing ApopTag Kits). The ApopTag product line provides various options in experimental design. A researcher can choose to detect staining by brightfield or fluorescence microscopy or by flow cytometry, depending on available expertise and equipment. There are also opportunities to study other proteins of interest in the context of apoptosis when using ApopTag Kits. By using antibodies conjugated with an enzyme other than peroxidase and an appropriate choice of substrate, it is possible to simultaneously examine another protein and apoptosis using ApopTag Peroxidase Kits.

Components

Equilibration Buffer 90416 3.0 mL -15°C to -25°C

Reaction Buffer 91417 2.0 mL -15°C to -25°C

TdT Enzyme 90418 0.672 mL -15°C to -25°C

Stop/Wash Buffer 90419 20 mL -15°C to -25°C

Anti-Digoxigenin-Peroxidase* 90420 3.0 mL 2°C to 8°C

Plastic Coverslips 90421 100 ea. Room Temp.

Control Slides 90422 2 each Room Temp.

DAB Substrate 90423 130 μL 2°C to 8°C

DAB Dilution Buffer 90424 6.5 mL 2°C to 8°C

*affinity purified sheep polyclonal antibody

Number of samples per kit: Sufficient materials are provided to stain 40 tissue specimens of approximately 5 cm2 each when used according to instructions. Reaction Buffer will be fully consumed before other reagents when kits are used for slide-mounted specimens.

Storage and Stability

Store the kit at -15°C to -25°C until the first use. After the first use, if the kit will be used within three months, store the TdT Enzyme (90418) at -15°C to -25°C and store the remaining components at 2°C to 8°C.

Precautions

1. The following kit components contain potassium cacodylate (dimethylarsinic acid) as a buffer: Equilibration Buffer (90416), Reaction Buffer (90417), and TdT Enzyme (90418). These components are harmful if swallowed; avoid contact with skin and eyes (wear gloves, glasses) and wash areas of contact immediately.

2. DAB (3,3′ diaminobenzidine) Substrate (90423) has been demonstrated to be a potential carcinogen and skin contact should be avoided. If skin contact does occur, flush with copious amounts of dH2O.

3. Antibody Conjugates (90420) contains 0.08% sodium azide as a preservative.

4. TdT Enzyme (90418) contains glycerol and will not freeze at -20°C. For maximum shelf life, do not warm this reagent to room temp. before dispensing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remzi Atilgan et al.
Turkish journal of medical sciences (2020-05-13)
To compare the effects of bilateral proximal tubal occlusion and bilateral total salpingectomy on ovarian reserve and the cholinergic system via rat experiment. Twenty - one adult female rats were randomly divided into the following three groups: G1 (n =
Effect of fluoride intoxication on endometrial apoptosis and lipid peroxidation in rats: role of vitamins E and C.
Mehmet Guney, Baha Oral, Gulnur Take, Seren Gulsen Giray, Tamer Mungan
Toxicology null
Heiko Sorg et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(9), 3049-3058 (2009-05-01)
The hematopoietic growth factor erythropoietin (EPO) attracts attention due to its all-tissue-protective pleiotropic properties. We studied the effect of EPO on dermal regeneration using intravital microscopy in a model of full dermal thickness wounds in the skin-fold chamber of hairless
Renal ischemia-reperfusion injury is prevented by the mineralocorticoid receptor blocker spironolactone.
Juan M Mejia-Vilet, Victoria Ramirez, Cristino Cruz, Norma Uribe, Gerardo Gamba, Norma A Bobadilla
American Journal of Physiology: Renal Physiology null
Cell-in-cell Structures Formed between Human Cancer Cell Lines and the Cytotoxic Regulatory T-cell Line HOZOT.
Takeuchi M, Inoue T, Otani T, Yamasaki F, Nakamura S, Kibata M
Journal of Molecular Cell Biology null

Articles

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service