Skip to Content
Merck
All Photos(2)

Key Documents

QGARDTXL04

Milli-Q

Q-Gard® Purification Cartridge

For use with Milli-Q® CLX/HX/HX SD/HR 7000 series systems, Produces high resistivity water from purification media.

Synonym(s):

Q-Gard® XL Polishing Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
NB.85
Pricing and availability is not currently available.

material

polypropylene and polyethylene cartridge (Virgin ion-exchange resins (for deionization))

Quality Level

feature

(Can produce high resistivity water from purification media.)

packaging

pkg of 1 unit

manufacturer/tradename

Q-Gard®

parameter

(Burst test: > 20 bar (> 290 psi))

capacity

32 g/L ion exchange capacity(Dynamic Ion Echange Capacity)
43 g/L total exchange capacity(NaCl)

compatibility

for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Q-Gard® XL Polishing Cartridge contains a combination of purification media carefully selected to remove trace ions to produce high-resistivity water.

The cartridge is constructed with clean and high-purity materials to minimize leaching of extractable contaminants.

For use with Milli-Q® CLX/HX/HX SD/HR 7000 series systems.

Application

This polishing cartridge contains virgin ion-exchange resins to remove trace ions and is recommended for any laboratory work requiring very low levels of ions from high volume systems.

Features and Benefits

  • Q-Gard® XL Purification Cartridge is tailored to the feed-water source to optimize the performance of downstream purification media to ensure consistent results.
  • e-Sure tag RFID technology for safety and automatic pack parameters archiving.
  • Replace every 6 months.
  • Genuine consumable manufactured in our ISO 9001-certified facility.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–9 of 9 Questions  
  1. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  2. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  3. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  4. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  5. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  6. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

  7. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  8. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

  9. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service