Select a Size
About This Item
Quality Segment
material
composite regenerated cellulose (CRC) membrane, polypropylene (Screen (feed)), polypropylene cap (end cap), polypropylene housing
product line
Pellicon®
storage condition
do not freeze
parameter
4-8 L/min-m2 flow rate
technique(s)
tangential flow filtration (TFF): suitable, ultrafiltration: suitable
H
0.89 cm (0.35 in.)
L
20.6 cm (8.1 in.)
filtration area
88 cm2
size
1.3 mL , Volume (feed), 2.8 mL , Volume (permeate)
width
5.6 cm (2.2 in.) , 56 mm
matrix
(Ultracel)
pore size
30 kDa pore size
pH
1-14
storage temp.
2-8°C
General description
filtration processes that require higher operating pressures, temperatures and
caustic cleaning regimes.
From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 design and automated manufacturing process provides unbeatable performance consistency between cassette sizes. Pellicon® 3 devices also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning.
Application
Features and Benefits
- Robust, void-free membrane provides optimum product retention and performance consistency
- Unique feed screen design enables high mass transfer, flux for higher final concentration
- Fast, reliable scale up/down from lab to production scale
- Automated manufacturing delivers unbeatable performance consistency and reliability
- Easy to install and clean
- Extreme temperature and chemical compatibility
Other Notes
Legal Information
Still not finding the right product?
Explore all of our products under Pellicon® 3 Cassette with Biomax® Membrane
Storage Class
10 - Combustible liquids
wgk
WGK 2
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.