Skip to Content

OP95

Anti-BRCA2 (Ab-1) Mouse mAb (2B)

liquid, clone 2B, Calbiochem®

Synonym(s):

Anti-Breast Cancer Susceptibility Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
2B, monoclonal
Species reactivity:
human
Application:
Citations:
129
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(Immunoblotting (2 µg/mL)
Immunoprecipitation (2 µg))

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRCA2(675)

General description

Anti-BRCA2 (Ab-1), mouse monoclonal, clone 2B, recognzies the ~460 kDa BRCA2 protein in cells expressing a wild type BRCA2 gene. It is validated for Western blotting and immunoprecipitation.
Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with myeloma cells. Recognizes the ~460 kDa BRCA2 protein.
Recognzies the ~460 kDa BRCA2 protein in cells expressing a wild type BRCA2 gene. Sold under license of U.S. Patent 5,753,441 and 6,162,897.

Immunogen

Human
a recombinant protein consisting of amino acids 1651-1821 of human BRCA2 fused to GST

Application

Immunoblotting (2 µg/ml)

Immunoprecipitation (2 µg)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In PBS.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
MCF7 cells

Other Notes

Andres, J.L., et al. 1998. Oncogene16, 2229.
Chen, J., et al. 1998. Mol. Cell2, 317.
Chen, P.L., et al. 1998. Proc. Natl. Acad. Sci. USA95, 5287.
Marmorstein, L.Y., et al. 1998. Proc. Natl. Acad. Sci. USA95, 13869.
Connor, F., et al. 1997. Nat. Genet.17, 423.
Sharan, S.K., et al. 1997. Nature386, 804.
Ludwig, T., et al. 1997. Genes Dev.11, 1226.
Mizuta, R., et al. 1997. Proc. Natl. Acad. Sci. USA94, 6927.
Rajan, J.V., et al. 1997. Dev. Biol.184, 385.
Suzuki, A., et al. 1997. Genes Dev.11, 1242.
Wong, A.K.C., et al. 1997. J. Biol. Chem.272, 31941.
Cannon-Albright, L.A. and M.H. Skolnick. 1996. Semin. Oncol.23, 1.
Goggins, M., et al. 1996. Cancer Res.56, 5360.
Stratton, M.R. 1996. Hum. Mol. Genet.5, 1515.
Tavtigian, S.V., et al. 1996. Nat. Genet.12, 333.
Rajan, J.V., et al. 1996. Proc. Natl. Acad. Sci. USA93, 13078.
Spillman, M.A. and A.M. Bowcock. 1996. Oncogene13, 1639.
Vaughn, J.P., et al. 1996. Cancer Res.56, 4590.
Wooster, R., et al. 1995. Nature378, 789.
The CAPAN-1 cell line, which has 6174delT mutation in the single BRCA2 allele, expresses a truncated BRCA2 protein of 230 kDa that contains the Anti-BRCA2 epitope and retains RAD51 binding but has lost p53 binding. Full length BRCA2 protein will co-migrate on SDS-PAGE with the catalytic subunit of DNA PK which can be detected using Anti-DNA PK, Cat. No. PC127. Antibody should be titrated for optimal results in individual systems.

Legal Information

Sold under license of U.S. Patents 5,837,492 and 6,124,104.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library