Skip to Content
Merck
All Photos(1)

Key Documents

OP93

Sigma-Aldrich

Anti-BRCA1 (Ab-2) Mouse mAb (MS13)

liquid, clone MS13, Calbiochem®

Synonym(s):

Anti-Breast Cancer Susceptibility Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MS13, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

General description

Anti-BRCA1 (Ab-2), mouse monoclonal, clone MS13, recognizes ~220 kDa BRCA1 in MCF-7 cells. It is validated for Western blotting, immunofluorescence, immunoprecipitation, and for paraffin sections.
Purified mouse monoclonal antibody generated by immunizing mice with the specified iummunogen and fusing splenocytes with NS1 mouse myeloma cells. Recognizes the ~220 kDa BRCA1 protein.
Recognizes the ~220 kDa BRCA1 protein in MCF-7 cells. Does not react with blood group antigens or growth factor receptors such as EGFR. Sold under license of U.S. Patent 5,753,441 and 6,162,897.

Immunogen

Epitope: within amino acids 1-304
Human
recombinant, human BRCA1

Application

Immunoblotting (2 µg/ml, chemiluminescence; see application references)

Immunofluorescence (see application references)

Immunoprecipitation (see application references)

Paraffin Sections (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Positive Control
MCF7 cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Scully, R., et al. 1996. Science272, 123.
Gudas, J.M., et al. 1996. Cell Growth Differ.7, 717.
Vaughn, J.P., et al. 1996. Cell Growth Differ.7, 711.
Goldgar, D.E. and Reilly, P.R. 1995. Nat. Genet.11, 113.
Merajver, S.D., et al. 1995. Clin. Can. Res.1, 539.
Merajver, S.D., et al. 1995. Nat. Genet.9, 439.
Struewing, J.P., et al. 1995. Nat. Genet.11, 198.
Thompson, M.E., et al. 1995. Nat. Genet.9, 444.
Futreal, P.A., et al. 1994. Science266, 120.
Miki, Y., et al. 1994. Science266, 66.

Legal Information

Sold under license of U.S. Patents 5,753,441 and 6,162,897.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Graziano et al.
The Journal of biological chemistry, 297(5), 101301-101301 (2021-10-15)
Lamin A/C provides a nuclear scaffold for compartmentalization of genome function that is important for genome integrity. Lamin A/C dysfunction is associated with cancer, aging, and degenerative diseases. The mechanisms whereby lamin A/C regulates genome stability remain poorly understood. We
Xiaolei Pan et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(29), E5940-E5949 (2017-07-05)
In the mammalian genome, certain genomic loci/regions pose greater challenges to the DNA replication machinery (i.e., the replisome) than others. Such known genomic loci/regions include centromeres, common fragile sites, subtelomeres, and telomeres. However, the detailed mechanism of how mammalian cells
S Fan et al.
Oncogene, 20(57), 8215-8235 (2002-01-10)
Unregulated expression of wild-type BRCA1 (wtBRCA1) confers an altered phenotype in cultured human prostate cancer cells, characterized by chemosensitivity, susceptibility to apoptosis, decreased DNA repair activity, and alterations of key cell regulatory proteins. We now report that the expression of
S Fan et al.
Oncogene, 20(35), 4827-4841 (2001-08-25)
The tumor suppressor activity of the BRCA1 gene product is due, in part, to functional interactions with other tumor suppressors, including p53 and the retinoblastoma (RB) protein. RB binding sites on BRCA1 were identified in the C-terminal BRCT domain (Yarden
Yong Xian Ma et al.
Oncogene, 22(1), 10-27 (2003-01-16)
The heat shock response is an evolutionarily conserved response to heat and other stresses that promotes the maintenance of key metabolic functions and cell survival. We report that exposure of human prostate (DU-145) and breast (MCF-7) cancer cells to heat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service