Select a Size
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
327, monoclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
mouse, human, rat
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
dilution
(Immunoblotting (2.5 µg/mL)
Immunoprecipitation (1 µg antibody)
Frozen Sections (not recommended)
Immunofluorescence (not recommended)
Paraffin Sections (not recommended))
isotype
IgG1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... SRC(6714)
Related Categories
General description
Immunogen
Application
Immunoprecipitation (1 µg antibody or use Cat. No. OP07A; see application references)
Frozen Sections (not recommended)
Immunofluorescence (not recommended)
Paraffin Sections (not recommended)
Packaging
Physical form
Analysis Note
NIH/3T3 cells
Other Notes
Cartwright, C.A., et al. 1985. Mol. Cell. Biol.5,, 2647.
Courtneidge, S.A. 1985. EMBO J.4, 1471.
Whitman, M., et al. 1985. Nature315, 239
Bolen, J.B., et al. 1984. Cell38, 767.
Hunter, T. and Sefton, B. 1980. Proc. Natl. Acad. Sci.77, 1311.
Collett, M.S. et al. 1979. Proc. Natl. Acad. Sci.76, 3159.
Collett, M.S. and Erikson, R.L. 1978. Proc. Natl. Acad. Sci.75, 2021.
Collett, M.S., et al. 1978. Cell15, 1363.
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| OP07-100UG | 04055977225006 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service