Skip to Content
Merck
All Photos(1)

Documents

OP03

Sigma-Aldrich

Anti-p53 (Ab-1) (Pantropic) Mouse mAb (PAb421)

liquid, clone PAb421, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb421, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rabbit, monkey, human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS1 mouse myeloma cells (see application references). Recognizes the ~53 kDa mutant and wild-type p53 proteins.
Recognizes the ~53 kDa mammalian wild-type and mutant p53 protein. Does not recognize phosphorylated or acetylated p53.
This Anti-p53 (Ab-1) (Pantropic) Mouse mAb (PAb421) is validated for use in Frozen Sections, Gel Shift, Immunoblotting, IF, FC, IP for the detection of p53 (Ab-1) (Pantropic).

Immunogen

Epitope: within amino acids 376-378 of human p53
Mouse
partially purified mouse p53

Application

Frozen Sections (10 µg/ml, see application references)

Gel Shift (see comments)

Immunoblotting (10 µg/ml, see application references)

Immunofluorescence (1-20 µg/ml, see application references)

Flow Cytometry (1-20 µg/ml)

Immunoprecipitation (1 μg per sample, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.

Analysis Note

Negative Control
SK-OV-3 cells or normal skin
Positive Control
Breast carcinoma, A431 cells, or p53 standard (Cat. No. 506147)

Other Notes

El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Legros, Y., et al. 1994. Oncogene9, 2071.
Barak, Y., et al. 1993. EMBO J.12, 461.
Kuerbitz, S.J. 1992. Proc. Natl. Acad. Sci. USA89, 7491.
Kastan, M.B., et al. 1992. Cell71, 587.
Lane, D.P. 1992. Nature358, 15.
Kastan, M.B., et al. 1991. Cancer Res.51 6304.
Crawford, L. and Harlow, E. 1982. J. Virol.41, 709.
Harlow, E., et al. 1981. J. Virol.39 861.
Originally called clone L21. Does not recognize phosphorylated p53. For a gel shift assay, use Cat. No. OP03L and resuspend in 100 µl buffer. Wild-type p53 has a short half-life and is present in low amounts in cells. For immunoprecipitation, increasing the amount of sample to be immunoprecipitated and applied to the gel may help visualize wild-type p53, and short incubation times with 35S-Met (≤1 h) will help reduce background. For immunoblotting of wild-type p53, maximize sensitivity by concentrating samples by immunoprecipitation with Cat. No. OP03, then immunoblot using PC35 and chemiluminescent detection. This antibody recognizes the region encompassing Ser376 to Ser378 (see application references) and does not recognize phosphorylated p53 (see application references). Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sun-Young Choi et al.
International journal of oncology, 40(1), 277-286 (2011-09-03)
Proteins involved in the G1 phase of the cell cycle are aberrantly expressed, sometimes in mutated forms, in human cancers including human hepatocellular carcinoma.
C J Chiou et al.
Journal of virology, 67(10), 6201-6214 (1993-10-01)
The 80-kDa IE2 nuclear phosphoprotein encoded by the human cytomegalovirus (HCMV) major immediate-early (MIE) gene behaves both as a nonspecific transactivator of heterologous reporter genes and as a specific repressor of its own promoter-enhancer region. To begin to examine the
M Schreiber et al.
Genes & development, 13(5), 607-619 (1999-03-11)
The c-jun proto-oncogene encodes a component of the mitogen-inducible immediate-early transcription factor AP-1 and has been implicated as a positive regulator of cell proliferation and G1-to-S-phase progression. Here we report that fibroblasts derived from c-jun-/- mouse fetuses exhibit a severe
Sricharan Bandhakavi et al.
Molecular & cellular proteomics : MCP, 9(2), 403-414 (2009-12-04)
Cellular nutritional and energy status regulates a wide range of nuclear processes important for cell growth, survival, and metabolic homeostasis. Mammalian target of rapamycin (mTOR) plays a key role in the cellular responses to nutrients. However, the nuclear processes governed
Jessica Q Ho et al.
PloS one, 6(2), e16815-e16815 (2011-02-25)
The pro-survival activity of NF-κB in response to a variety of stimuli has been extensively characterized. Although there have been a few reports addressing the pro-cell death role of NF-κB, the precise mechanism of NF-κB's pro-cell death function still remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service