Skip to Content
Merck

Skip To

MX0SP20CL3

Millistak+® HC Pro Depth Filter in Micro 20 format

X0SP, filtration area 20 cm², Process Development Scale

greener alternative

Greener Alternative

Synonym(s):

20 cm2 surface area, Female-Luer fitting connection, Millistak+ HC Pro Pod Depth Filter, Millistak+ X0SP Micro 20 Depth Filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23151806
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Millistak+® HC Pro Depth Filter in Micro 20 format, X0SP, silica filter aid (with Polyacrylic fiber), filtration area 20 cm2, hydrophilic

Quality Segment

material

glass filled polypropylene housing, silica filter aid (with Polyacrylic fiber)

feature

hydrophilic

manufacturer/tradename

Millistak+®

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

2.5 in. (6.4 cm)

filtration area

20 cm2

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

input

sample type liquid

fitting

Luer fitting

filter grade

X0SP

greener alternative category

shipped in

ambient

General description

Device Configuration: Micro 20
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

Secondary clarification (direct harvest and centrate), and downstream filtration

Features and Benefits

Millistak+® HC Pro Micro 20 devices offer:

  • Improved scalability during media screening trials
  • Smaller filtration area slightly reduces feedstream volume requirements (approximately 15% less).
  • Transparent housing enables visibility of fluid flow
  • Easier to calculate flow rates based on 20 cm2 filter area


Millistak+®HC Pro media offers:
  • Synthetic materials of construction
  • Reduced TOC extractables and a 50% reduction in the recommended pre-use flush volumes
  • No beta glucans to interfere with limulous amoebocyte lysate (LAL) testing for bacterial endotoxins
  • Lot-to-lot consistency for successful development and implementation of robust clarification processes
  • Two times the filtration capacity with equivalent filter retention properties over commercial DE-based benchmarks
  • Improved HCP impurity clearance


Millistak+® Pod filters offer:
  • Low hold-up volume for greater product yield
  • Flexible, modular format offers scalability up to 20,000 liters
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Packaging

3 EA

Other Notes

Integrity is maintained after 2 autoclave cycles of 60 minutes at 123 °C

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

MILLISTAK+® is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MC0SP20CL3MD0SP20CL3MC0SP014H1
description

X0SP, silica filter aid (with Polyacrylic fiber), filtration area 20 cm2, hydrophilic

description

C0SP, silica filter aid (with Polyacrylic fiber), filtration area 20 cm2, hydrophilic

description

D0SP, silica filter aid (with Polyacrylic fiber/ non-woven material), filtration area 20 cm2, hydrophilic

description

silica filter aid, acrylic, filtration area 135 cm2, hydrophilic

fitting

Luer fitting

fitting

Luer fitting

fitting

Luer fitting

fitting

-

filter grade

X0SP

filter grade

C0SP

filter grade

D0SP

filter grade

C0SP

filtration area

20 cm2

filtration area

20 cm2

filtration area

20 cm2

filtration area

135 cm2

material

silica filter aid (with Polyacrylic fiber), glass filled polypropylene housing

material

glass filled polypropylene housing, silica filter aid (with Polyacrylic fiber)

material

glass filled polypropylene housing, silica filter aid (with Polyacrylic fiber/ non-woven material)

material

polypropylene , acrylic , silica filter aid

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters