Skip to Content
Merck
All Photos(1)

Key Documents

MX0487

Supelco

Methanol

≥99.8% (GC), anhydrous, reagent grade, ACS reagent, for extraction, for analytical purposes, meets USP/NF

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
41116121
EC Index Number:
200-659-6
grade:
ACS reagent
anhydrous
for extraction
reagent grade
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

grade

ACS reagent
anhydrous
for extraction
reagent grade

Quality Level

Agency

meets USP/NF

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

for analytical purposes

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Titrable base
≤0.0003 meq/g Titrable acid
≤0.001% Carbonyl (acetone, formaldehyde, acetaldehyde)
≤0.03% Water (H2O)

evapn. residue

≤0.001%

color

clearAPHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

AppearanceClearAssay (GC)99.8% minCarbonyl (acetone,formaldehyde,acetaldehyde)0.001% maxColor (APHA)10 maxEvaporation residue0.001% maxSolubility in waterTo pass testSubstances darkened by sulfuric acidTo pass testSubstances reducing permanganateTo pass testTitrable acid0.0003meq/g maxTitrable base0.0002meq/g maxWater (H2O)0.03% max
Evaporation Number: 1.9

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service