Skip to Content
Merck

Skip To

MSIPS4W10

Multiscreen® 96 well Plate, hydrophobic PVDF membrane

pore size 0.45 μm, sterile

Multiscreen® 96 well Plate, hydrophobic PVDF membrane

Synonym(s):

Hydrophobic 96-Well Plate, PVDF 96-Well Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Material:
Durapore PVDF membrane, flat bottom wells, white acrylic plate
Pore size:
0.45 μm pore size
Sterility:
sterile
Binding type:
high binding surface
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

Durapore PVDF membrane, flat bottom wells, white acrylic plate

description

Sterile, white 96-well filter plate with 0.45 um pore size Hydrophobic PVDF membrane for ELISPOT Assays. Comes in a pack of 10.

sterility

sterile

usage

Automated

feature

hydrophobic, lid

packaging

pkg of 10 plates

manufacturer/tradename

MultiScreen®

technique(s)

ELISPOT: suitable, cell based assay: suitable, dot blot: suitable

H

14.4 mm

L

127.8 mm

W

85.5 mm

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

matrix

Immobilon®-P

pore size

0.45 μm pore size

binding type

high binding surface

compatibility

centrifuge & manifold

shipped in

ambient

storage temp.

10-30°C

General description

The polyvinylidene fluoride (PVDF) membrane plates are recommended, over a mixed cellulose ester format, due to slightly improved binding of capture antibody and superior performance in spot detection, particularly for fluorescent applications.

Application

MultiScreen® HTS IP Filter Plate, 0.45 μm, white, sterile has been used in enzyme-linked immunospot (ELISpot) assay using:
  • dsDNA-producing plasma cells[1]
  • memory B cells and bone marrow localized long-lived plasma cells[2]
  • draining lymph node and spleen cells

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSHVN4BMAIPN4550MSIPN4B10
pore size

0.45 μm pore size

pore size

0.45 μm pore size

pore size

0.45 μm pore size

pore size

0.45 μm pore size

sterility

sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

material

Durapore PVDF membrane, white acrylic plate, flat bottom wells

material

Durapore PVDF membrane, flat bottom wells, white Barex® plate (co-polymer)

material

Durapore PVDF membrane, clear acrylic plate, flat bottom wells

material

Durapore PVDF membrane, flat bottom wells, white Barex® plate (co-polymer)

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

manufacturer/tradename

MultiScreen®

binding type

high binding surface

binding type

low binding surface

binding type

high binding surface

binding type

high binding surface

feature

hydrophobic, lid

feature

hydrophilic, lid

feature

hydrophobic, lid

feature

hydrophobic, lid


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Do MSIPS4W10 and S5EM077I10 refer to the same plate?

    1 answer
    1. Yes, both MSIPS4W10 and S5EM077I10 correspond to the same plate, identified by the description: MultiScreen HTS IP PVDF 0.45um White Sterile 10pk. MSIPS4W10 is the public catalog number, whereas S5EM077I10 is a custom order request for a specific customer.

      Helpful?

Reviews

Active Filters

  1. Boston, MA
    • Reviews 2
    • Votes 2
    5 out of 5 stars.

    Great plate for ELISPOTS

    These plates give a strong ELISPOT signal with very low background and work well with either a vacuum manifold plate washing apparatus or manual washing.

    Helpful?