Select a Size
About This Item
Quality Segment
material
flat bottom wells, Durapore PVDF membrane, clear acrylic plate
description
Sterile, clear 96-well filter plate with 0.45 um pore size Hydrophobic PVDF membrane for ELISPOT Assays. Comes in a pack of 10.
sterility
sterile
product line
MultiScreen®
usage
Automated
feature
hydrophobic, lid
packaging
pkg of 10 plates
technique(s)
ELISPOT: suitable, cell based assay: suitable, dot blot: suitable
H
14.4 mm
L
127.8 mm
W
85.5 mm
filtration area
0.28 cm2
plate size
96 wells
well maximum volume
300 μL
working volume
50-250 μL
matrix
Immobilon®-P
pore size
0.45 μm pore size
input
sample type hematopoietic stem cell(s)
sample type pancreatic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type induced pluripotent stem cell(s)
sample type mesenchymal stem cell(s)
sample type: human embryonic stem cell(s)
sample type neural stem cell(s)
sample type cardiac stem cell(s)
binding type
high binding surface
compatibility
centrifuge & manifold
shipped in
ambient
storage temp.
10-30°C
General description
HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.
- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.
Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.
Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.
Application
Assays
Cell Culture
Physical form
Other Notes
Legal Information
Still not finding the right product?
Explore all of our products under Multiscreen® 96 well Plate, hydrophobic PVDF membrane
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.