Skip to Content
Merck

Skip To

MP0DADAPT

Disposable Adapter Kit 3 through adapters, 3 blind adapters

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23151806
NACRES:
NB.24
eCl@ss:
32031690
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

ambient

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MP0DADPTFXX42PM001CAD6163
shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

-

Quality Level

400

Quality Level

400

Quality Level

SP1

Quality Level

-




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters