Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.79
UNSPSC Code:
41116129
packaging
pack of 1 kit
Quality Segment
manufacturer/tradename
Milliflex Oasis®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis® pumps
shipped in
ambient
General description
MILLIFLEX OASIS® sanitization head enables to sanitize the complete pump at once. The sanitization head can be autoclaved at 121°C for up to 30 min or at 134°C for up to 15 min. This sanitization head is a walk-away system to clean efficiently the interior of the MILLIFLEX OASIS® pump.
Application
MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis
Legal Information
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under MILLIFLEX OASIS® flow sanitization kit
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MMSANKIT1 | 04061839792821 |