Skip to Content
Merck
All Photos(2)

Documents

MMMP1MAG-79K

Millipore

MILLIPLEX® Mouse MMP Magnetic Bead Panel 1

TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in mouse serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

standard curve range: 12-50,000 pg/mL
(MMP-8)

standard curve range: 2-10,000 pg/mL
(MMP-3)

standard curve range: 49-200,000 pg/mL
(MMP-2)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

MILLIPLEX® Mouse MMP Bead Panel 1 is a 3 plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum/plasma samples: MMP-2, MMP-3 and MMP-8.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: MMP-2, MMP-3, MMP-8
  • Recommended Sample type: serum or plasma
  • Recommended Sample dilution: 1:100
  • Assay Run Time: Overnight or one day
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Other Notes

Sensitivity: See protocol for sensitivities of individual analytes

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clélia Le Gallic et al.
PloS one, 10(6), e0128539-e0128539 (2015-06-06)
After Chernobyl and Fukushima Daï Chi, two major nuclear accidents, large amounts of radionuclides were released in the environment, mostly caesium 137 (137Cs). Populations living in contaminated territories are chronically exposed to radionuclides by ingestion of contaminated food. However, questions
Adrian Filiberti et al.
Investigative ophthalmology & visual science, 61(10), 24-24 (2020-08-14)
Corneal opacity and neovascularization (NV) are often described as outcomes of severe herpes simplex virus type 1 (HSV-1) infection. The current study investigated the role of colony-stimulating factor 1 receptor (CSF1R)+ cells and soluble factors in the progression of HSV-1-induced
Fang Han et al.
Investigative ophthalmology & visual science, 61(14), 24-24 (2020-12-22)
The purpose of this study was to explore the role of thymic stromal lymphopoietin (TSLP) secreted by Aspergillus fumigatus-stimulated dendritic cells (DCs) during the T helper 17 (Th17) immune response, and further clarify the mechanisms contributing to the Th17 immune

Related Content

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service