MKI2MAG-94K
MILLIPLEX® Mouse Kidney Injury Magnetic Bead Panel 2 - Toxicity Multiplex Assay
The analytes available for this multiplex kit are: Clusterin, Cystatin C, EGF, Lipocalin-2/NGAL, Osteopontin (OPN) (for urine samples) or Clusterin, Cystatin C, Lipocalin-2/NGAL, Osteopontin (OPN) (for serum/plasma samples).
About This Item
Recommended Products
Quality Level
species reactivity
mouse
manufacturer/tradename
Milliplex®
assay range
accuracy: 90-99%
sensitivity: 0.005-0.190 ng/mL
(MinDC+2SD)
standard curve range: 0.005-5 ng/mL
(Lipocalin-2/NGAL)
standard curve range: 0.01-10 ng/mL
(Osteopontin (OPN))
standard curve range: 0.04-40 ng/mL
(EGF)
standard curve range: 0.05-50 ng/mL
(Cystatin C)
standard curve range: 0.2-250 ng/mL
(Clusterin)
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex xMAP)
shipped in
wet ice
General description
The MILLIPLEX® Mouse Kidney Injury Bead Panel 2 contains all the components necessary to measure the following five biomarkers in any combination using Luminex® xMAP® technology: Clusterin, Cystatin C, EGF, Lipocalin-2/NGAL, Osteopontin (OPN). The kit uses a 96-well format, contains a lyophilized standard cocktail, two quality controls and can measure up to 38 urine samples in duplicate.
Panel Type: Toxicity
Specificity
Application
- Analytes: Clusterin, Cystatin C, EGF, Lipocalin-2/NGAL, Osteopontin (OPN)
- Recommended Sample type: urine
- Recommended Sample dilution: 1:1,000
- Assay Run Time: overnight
- Research Category: Toxicity
Features and Benefits
Packaging
Storage and Stability
Other Notes
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.
Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.
Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.
Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service