Select a Size
About This Item
form
powder (dehydrated)
shelf life
60 mo.
packaging
pkg of 110 g
color
light green-beige dehydrated powder, clear light purpleslightly opalescent prepared medium
input
sample type food(s)
sample type sport drink
sample type wine
sample type soft drink
sample type cider
sample type cosmetics
sample type bottled water
sample type beer
sample type beverage(s)
pH
7.2±0.2 (25.0 °C)
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
microbiology
water monitoring
compatibility
for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®
shipped in
ambient
storage temp.
2-30°C
suitability
Aspergillus niger, Bacillus subtilis, Candida albicans, Clostridium sporogenes, Pseudomonas aeruginosa, Staphylococcus aureus
Preparation Note
Analysis Note
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MB000000S | 04053252250330 |
